Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
LSR has a probable role in the clearance of triglyceride-rich lipoprotein from blood. It binds chylomicrons, LDL and VLDL in presence of free fatty acids and allows their subsequent uptake in the cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Angulin-1; immunoglobulin-like domain containing receptor 3; Lipolysis-stimulated lipoprotein receptor; lipolysis-stimulated remnant; LISCH protein; liver-specific bHLH-Zip transcription factor
Gene Aliases: ILDR3; LISCH; LISCH7; LSR
UniProt ID: (Human) Q9UQL3
Entrez Gene ID: (Human) 51599
Molecular Function:
defense/immunity protein
immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support