Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
BLTR2 has been reported in humans in peripheral blood leukocytes, mononuclear lymphocytes, and spleen. ESTs have been isolated from a broad array of human libraries, including brain, eye, fetal lung/testis/B-cell, heart, kidney, melanocyte/uterus/fetal heart, testis, tonsil, and uterus libraries.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Blt-2; Hblt2; Leukotriene B4 receptor 2; Leukotriene B4 receptor BLT2; LTB4 receptor JULF2; LTB4-R 2; LTB4-R2; Seven transmembrane receptor BLTR2
Gene Aliases: 5830462O07Rik; BLT2; BLT2R; BLTR2; JULF2; KPG_004; LTB4-R 2; LTB4-R-2; LTB4-R2; LTB4R2; NOP9
UniProt ID: (Human) Q9NPC1, (Rat) Q924U0, (Mouse) Q9JJL9
Entrez Gene ID: (Human) 56413, (Rat) 114098, (Mouse) 57260
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support