Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene belongs to the family of latent TGF-beta binding proteins. The secretion and activation of TGF-betas is regulated by their association with latency-associated proteins and with latent TGF-beta binding proteins. The product of this gene targets latent complexes of transforming growth factor beta to the extracellular matrix, where the latent cytokine is subsequently activated by several different mechanisms. Alternatively spliced transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LanC (bacterial lantibiotic synthetase component C)-like 1; latent TGF beta binding protein; Latent-transforming growth factor beta-binding protein 1; LTBP-1; MGC163161; TGF-beta-1-BP-1; TGF-beta1-BP-1; Transforming growth factor beta-1-binding protein 1; Transforming growth factor beta-1-masking protein large subunit; transforming growth factor-beta (TGF-beta) masking protein large subunit
Gene Aliases: 9430031G15Rik; 9830146M04; b2b1000Clo; Ltbp-1; LTBP1; Tgfb
UniProt ID: (Human) Q14766, (Mouse) Q8CG19, (Rat) Q00918
Entrez Gene ID: (Human) 4052, (Mouse) 268977, (Rat) 59107
Molecular Function:
extracellular matrix structural protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support