Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MRTALLLLAT LAVATGPALT LRCHVCTSSS NCKHSVVCPA SSRFCKTTNT VEPLRGNLVK KDCAESCTPS YTLQGQVSSG TSSTQCCQED LCNEKLHNAA PT
May act as a specification marker at earliest stage specification of lymphocytes between B- and T-cell development. Marks the earliest stage of B-cell specification. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E48 antigen; Ly-6D; Lymphocyte antigen 6D
Gene Aliases: E48; Ly-6D; LY6D
UniProt ID: (Human) Q14210
Entrez Gene ID: (Human) 8581
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support