Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 3
Immunogen sequence: TDEGDNRVWC HVCERENTFE CQNPRRCKWT EPYCVIAAVK IFPRFFMVAK QCSAGCAAME RPKPEEKRFL LEEPMPFFYL KCCKIRYCNL EGPPINSSVF KEYAG
Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 36%.
Binds to an epitope located within the peptide sequence LEEPMPFFYLKCCKI as determined by overlapping synthetic peptides.
Required for sperm migration into the oviduct and male fertility by controlling binding of sperm to zona pellucida. May play a role in cell growth (PubMed:18089789). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cancer/testis antigen 97; FLJ35226; Ly-6K; Lymphocyte antigen 6K; up-regulated in lung cancer 10
Gene Aliases: CO16; CT97; HSJ001348; ly-6K; LY6K; URLC10
UniProt ID: (Human) Q17RY6
Entrez Gene ID: (Human) 54742
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support