Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: HepG2.
Predicted reactivity: Mouse (88%), Rat (87%), Dog (89%), Rhesus Monkey (97%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Lysophospholipases are enzymes that act on biological membranes to regulate the multifunctional lysophospholipids. The protein encoded by this gene hydrolyzes lysophosphatidylcholine to glycerophosphorylcholine and a free fatty acid. This enzyme is present in the plasma and thought to be associated with high-density lipoprotein. A later paper contradicts the function of this gene. It demonstrates that this gene encodes a lysosomal enzyme instead of a lysophospholipase and has both calcium-independent phospholipase A2 and transacylase activities.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-O-acylceramide synthase; ACS; LCAT-like lysophospholipase; LLPL; LPLA2; Lysophospholipase 3; lysophospholipase 3 (lysosomal phospholipase A2); Lysosomal phospholipase A and acyltransferase; Lysosomal phospholipase A2; Phospholipase A2 group XV; phospholipase A2, group XV
Gene Aliases: ACS; GXVPLA2; LLPL; LPLA2; LYPLA3; PLA2G15; UNQ341/PRO540
UniProt ID: (Human) Q8NCC3
Entrez Gene ID: (Human) 23659
Molecular Function:
acyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support