Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
LYSMD3 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LysM and putative peptidoglycan-binding domain-containing protein 3; LysM, putative peptidoglycan-binding, domain containing 3
Gene Aliases: 1110030H10Rik; BC003322; LYSMD3
UniProt ID: (Human) Q7Z3D4, (Mouse) Q99LE3
Entrez Gene ID: (Human) 116068, (Mouse) 80289
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support