Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: VQEQLHMAEK ELEKKFQQTA AYRNMKEILT KKNDQIKDLR KRLAQYEPED
Sequence homology: Cow: 100%; Dog: 86%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 92%
The function remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: leucine zipper transcription factor-like 1; Leucine zipper transcription factor-like protein 1
Gene Aliases: BBS17; LZTFL1
UniProt ID: (Human) Q9NQ48
Entrez Gene ID: (Human) 54585
Molecular Function:
basic leucine zipper transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support