Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: EPLCPAVPPR KAVPVTSFTY INEDFRTESP PSPSSDVEDA REQRAHNAHL
Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%
LZTS2 is a negative regulator of katanin-mediated microtubule severing and release from the centrosome. LZTS2 is required for central spindle formation and the completion of cytokinesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hLZTS2; Leucine zipper putative tumor suppressor 2; leucine zipper, putative tumor suppressor 2; Protein LAPSER1; RP11-108L7.8
Gene Aliases: KIAA1813; LAPSER1; LZTS2
UniProt ID: (Human) Q9BRK4
Entrez Gene ID: (Human) 84445
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support