Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Lactate dehydrogenase C catalyzes the conversion of L-lactate and NAD to pyruvate and NADH in the final step of anaerobic glycolysis. LDHC is testis-specific and belongs to the lactate dehydrogenase family. Two transcript variants have been detected which differ in the 5' untranslated region.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer/testis antigen 32; Cell proliferation-inducing gene 19 protein; CT32; epididymis secretory sperm binding protein Li 133P; L-lactate dehydrogenase A chain; L-lactate dehydrogenase C chain; lactate dehydrogenase 1, A chain; lactate dehydrogenase 3, C chain, sperm specific; lactate dehydrogenase A-like; lactate dehydrogenase A4; lactate dehydrogenase c variant 1; lactate dehydrogenase C variant 3; lactate dehydrogenase c variant 4; lactate dehydrogenase c variant 5; lactate dehydrogenase C4; lactate dehydrogenase M; lactate dehydrogenase-A; lactate dehydrogenase-X; LDH muscle subunit; LDH testis subunit; LDH-A; LDH-C; LDH-M; LDH-X; M-LDH; proliferation-inducing gene 19; Renal carcinoma antigen NY-REN-59; testis secretory sperm-binding protein Li 234P
Gene Aliases: CT32; GSD11; HEL-S-133P; l7R2; Ldh-1; Ldh-3; LDH-C4; Ldh-x; Ldh1; LDH3; LDHA; LDHC; Ldhc4; LDHM; LDHX; PIG19
UniProt ID: (Human) P00338, (Human) P07864, (Rat) P04642, (Mouse) P06151, (Mouse) P00342
Entrez Gene ID: (Human) 3939, (Human) 3948, (Rat) 24533, (Mouse) 16828, (Rat) 29634, (Mouse) 16833
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support