Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For ICC, formaldehyde fixation is recommended. Permeabilization with Triton-X 100 is recommended for formaldehyde-fixed cells. For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Based on 100% sequence identity, this antibody is predicted to react with Pig
Lamins are a class of intermediate filament proteins that form a matrix on the inner surface of the nuclear envelope. These proteins are found in many different cell types in three different forms (A, B, and C). Lamins A and C are alternatively spliced versions of the LMNA gene. The LMNA gene has been linked to many disorders of the muscular system, nervous system, and the fat distributions systems including: Emery-Dreifuss muscular dystrophy, Dunnigan-type familial partial lipodystrophy (FPLD), limb-girdle muscular dystrophy (LGMD1B), dilated cardiomyopathy (CMD1A), axonal neuropathy (Charcot-Marie-Tooth disease; CMT2B1), and mandibuloacral dysplasia (MAD).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 70 kDa lamin; Lamin A+C mutant; lamin a-c; lamin A/C-like 1; Lamin AC; lamin C; lamin-A/C; mutant 453W; Mutant lamin A/C; Prelamin-A/C; progerin mutant; renal carcinoma antigen NY-REN-32; RP11-54H19.1
Gene Aliases: CDCD1; CDDC; CMD1A; CMT2B1; Dhe; EMD2; FPL; FPLD; FPLD2; HGPS; IDC; LDP1; LFP; LGMD1B; LMN1; LMNA; LMNC; LMNL1; PRO1
UniProt ID: (Human) P02545, (Mouse) P48678
Entrez Gene ID: (Human) 4000, (Mouse) 16905
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support