Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
Antibody detects endogenous levels of total Laminin 2 alpha.
Laminin antibodies are widely used to label blood vessels and basement membranes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dystrophia muscularis; Laminin A chain; Laminin B1 chain; laminin B1 subunit 1; Laminin B2 chain; Laminin M chain; Laminin subunit alpha-1; Laminin subunit alpha-2; Laminin subunit beta-1; Laminin subunit gamma-1; laminin, alpha 1; laminin, alpha 2; laminin, beta 1; laminin, gamma 1 (formerly LAMB2); laminin-1; Laminin-1 subunit alpha; Laminin-1 subunit beta; Laminin-1 subunit gamma; Laminin-10 subunit beta; Laminin-10 subunit gamma; Laminin-11 subunit gamma; Laminin-12 subunit alpha; Laminin-12 subunit beta; Laminin-2 subunit alpha; Laminin-2 subunit beta; Laminin-2 subunit gamma; Laminin-3 subunit alpha; Laminin-3 subunit gamma; Laminin-4 subunit alpha; Laminin-4 subunit gamma; Laminin-6 subunit beta; Laminin-6 subunit gamma; Laminin-7 subunit gamma; Laminin-8 subunit beta; Laminin-8 subunit gamma; Laminin-9 subunit gamma; Merosin heavy chain; S-LAM alpha; S-LAM gamma; S-laminin subunit alpha; S-laminin subunit gamma
Gene Aliases: 5830440B04; AA408497; B2e; C77966; C80098; C81607; CLM; D130003D08Rik; dy; LAMA; Lama-1; LAMA1; LAMA2; Lamb-1; LAMB1; Lamb1-1; LAMB2; LAMC1; LAMM; LIS5; mer; merosin; mKIAA4087; PTBHS; S-LAM-alpha
UniProt ID: (Human) P25391, (Human) P24043, (Human) P07942, (Human) P11047, (Mouse) P19137, (Mouse) Q60675, (Mouse) P02469
Entrez Gene ID: (Human) 284217, (Human) 3908, (Human) 3912, (Human) 3915, (Mouse) 16772, (Rat) 316758, (Rat) 309368, (Mouse) 16773, (Mouse) 16777, (Rat) 298941, (Mouse) 226519, (Rat) 117036
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support