Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • Laminin Antibodies

            AbboMax

            Laminin Monoclonal Antibody (ABM412)

            View all (17) Laminin antibodies
            Datasheet
            Protocols
            Questions & Answers
            Catalog # 500-7794
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            323.00
            100 µg
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite Laminin Monoclonal Antibody (ABM412)

            • Antibody Testing Data (1)
            Application
            Laminin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 1

            Laminin Antibody (500-7794) in IHC (P)

            Immunohistochemistry: Human lung tissue FFPE slides stained with mouse anti-Laminin monoclonal antibody (Cat#500-7794) at 1:200 for 10 min @ RT, Visualized by using DAB chromogen color system. Note: Staining of formalin-fixed tissue requires boiling tissue sections in 10 mM Citrate Buffer, pH 6.0 for 10 min followed by cooling at RT for 20 min. H.
            View Product

            Laminin Antibody in Immunohistochemistry (Paraffin) (IHC (P))

            Product Details

            500-7794

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            0.1-1 µg/mL
            -

            Immunohistochemistry (Paraffin) (IHC (P))

            2-10 µg/mL
            -
            Product Specifications

            Species Reactivity

            Bovine, Chicken, Human, Rat

            Host/Isotype

            Mouse / IgG2b

            Class

            Monoclonal

            Type

            Antibody

            Clone

            ABM412

            Immunogen

            Purified recombinant partial length of human Laminin protein

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.5 mg/mL

            Purification

            Protein A

            Storage buffer

            PBS with proprietary stabilizer

            Contains

            0.01% sodium azide

            Storage conditions

            Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            Product Specific Information

            Positive control: Lung, heart

            Cellular location: Extracellular matrix, Cytoplasmic.

            Target Information

            Laminin antibodies are widely used to label blood vessels and basement membranes.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: extra-cellular matrix protein; Laminin A chain; laminin B1; Laminin B1 chain; laminin B1 subunit 1; Laminin B2 chain; laminin beta-1-1 chain; Laminin M chain; Laminin subunit alpha-1; Laminin subunit alpha-2; Laminin subunit beta-1; Laminin subunit gamma-1; laminin, alpha 1; laminin, alpha 2; laminin, alpha 2 (merosin, congenital muscular dystrophy); laminin, beta 1; laminin, gamma 1 (formerly LAMB2); laminin-1; Laminin-1 subunit alpha; Laminin-1 subunit beta; Laminin-1 subunit gamma; Laminin-10 subunit beta; Laminin-10 subunit gamma; Laminin-11 subunit gamma; Laminin-12 subunit alpha; Laminin-12 subunit beta; Laminin-2 subunit alpha; Laminin-2 subunit beta; Laminin-2 subunit gamma; Laminin-3 subunit alpha; Laminin-3 subunit gamma; Laminin-4 subunit alpha; Laminin-4 subunit gamma; Laminin-6 subunit beta; Laminin-6 subunit gamma; Laminin-7 subunit gamma; Laminin-8 subunit beta; Laminin-8 subunit gamma; Laminin-9 subunit gamma; Merosin heavy chain; S-LAM alpha; S-LAM gamma; S-laminin subunit alpha; S-laminin subunit gamma

            View more View less

            Gene Aliases: B2e; CLM; LAMA; LAMA1; LAMA2; LAMB1; Lamb1-1; LAMB2; LAMC1; LAMM; LIS5; PTBHS; S-LAM-alpha

            View more View less

            UniProt ID: (Human) P25391, (Human) P24043, (Human) P07942, (Human) P11047

            View more View less

            Entrez Gene ID: (Chicken) 374016, (Human) 284217, (Chicken) 421709, (Human) 3908, (Human) 3912, (Chicken) 396478, (Bovine) 520030, (Human) 3915, (Bovine) 532572, (Chicken) 424442, (Rat) 316758, (Rat) 309368, (Rat) 298941, (Rat) 117036

            View more View less

            Function(s)

            receptor binding extracellular matrix structural constituent protein binding structural molecule activity integrin binding molecular_function glycosphingolipid binding enzyme binding extracellular matrix protein

            Process(es)

            cell adhesion cell surface receptor signaling pathway regulation of cell adhesion extracellular matrix organization regulation of cell migration regulation of embryonic development axon guidance muscle organ development myelination in peripheral nervous system positive regulation of synaptic transmission, cholinergic neuronal-glial interaction involved in cerebral cortex radial glia guided migration positive regulation of cell migration neuron projection development substrate adhesion-dependent cell spreading endodermal cell differentiation odontogenesis positive regulation of epithelial cell proliferation negative regulation of cell adhesion organ morphogenesis tissue development cell migration basement membrane assembly protein complex assembly endoderm development extracellular matrix disassembly hemidesmosome assembly morphogenesis of an epithelial sheet biological_process establishment of epithelial cell apical/basal polarity retina development in camera-type eye epithelial tube branching involved in lung morphogenesis branching involved in salivary gland morphogenesis retinal blood vessel morphogenesis embryo implantation
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-574q8:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0