Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AA408497; Lama; Laminin B1s chain; Laminin B2 chain; Laminin chain B3; laminin chain beta 2; Laminin S; Laminin subunit beta-2; Laminin subunit gamma-1; laminin, beta 2 (laminin S); laminin, gamma 1 (formerly LAMB2); Laminin-1 subunit gamma; Laminin-10 subunit gamma; Laminin-11 subunit beta; Laminin-11 subunit gamma; Laminin-14 subunit beta; Laminin-15 subunit beta; Laminin-2 subunit gamma; Laminin-3 subunit beta; Laminin-3 subunit gamma; Laminin-4 subunit beta; Laminin-4 subunit gamma; Laminin-6 subunit gamma; Laminin-7 subunit beta; Laminin-7 subunit gamma; Laminin-8 subunit gamma; Laminin-9 subunit beta; Laminin-9 subunit gamma; S-LAM beta; S-LAM gamma; S-laminin subunit beta; S-laminin subunit gamma
Gene Aliases: AW211941; B2e; Lamb-2; LAMB2; LAMC1; LAMS; NPHS5; npht; SLAM
UniProt ID: (Human) P55268, (Human) P11047, (Mouse) Q61292, (Rat) P15800
Entrez Gene ID: (Human) 3913, (Human) 3915, (Mouse) 16779, (Rat) 25473, (Mouse) 226519, (Rat) 117036
Molecular Function: extracellular matrix protein DNA metabolism protein chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support