Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Recommended positive controls: A431, NIH-3T3, mouse bone marrow.
Predicted reactivity: Mouse (81%), Rat (80%), Bovine (80%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The product encoded by this gene is a laminin that belongs to a family of basement membrane proteins. This protein is a beta subunit laminin, which together with an alpha and a gamma subunit, forms laminin-5. Mutations in this gene cause epidermolysis bullosa junctional Herlitz type, and generalized atrophic benign epidermolysis bullosa, diseases that are characterized by blistering of the skin. Multiple alternatively spliced transcript variants that encode the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Epiligrin subunit bata; Kalinin B1 chain; Kalinin subunit beta; kalinin-140kDa; Laminin B1k chain; laminin beta 3; laminin S B3 chain; Laminin subunit beta-3; laminin, beta 3 (nicein (125kD), kalinin (140kD), BM600 (125kD)); Laminin-5 subunit beta; Nicein subunit beta; nicein, 125kD; nicein, 125kDa; nicein-125k; nicein-125kDa; RP1-272L16.3
Gene Aliases: AI1A; BM600-125KDA; LAM5; LAMB3; LAMNB1
UniProt ID: (Human) Q13751, (Mouse) Q61087
Entrez Gene ID: (Human) 3914, (Mouse) 16780
Molecular Function: extracellular matrix protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support