Search
Search
Invitrogen
Immunogen sequence: CLPCNCSGRS EECTFDRELF RSTGHGGRCH HCRDHTAGPH CERCQENFYH WDPRMPCQPC DCQSAGSLHL QCDDTGTCAC KPTVTGWKCD RCLPGFHSLS EGGCRPCTCN PAGSLDTCDP RSGRCPCKEN VEG
Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 83%.
Laminin is a non-collagenous protein of basement membranes. By rotary shadowing it has a cross-like shape, consisting of one long arm, (400 kD) and three short arms (200 kD). Antibodies to laminin localize exclusively in basement membranes. Laminin is found throughout the basement membrane, but the reaction with the antibodies to laminin is more intense in the lamina lucida, the part of the basement membrane adjacent to the cell membrane. Laminin binds to type IV collagen, a component exclusively localized in the basement membrane. Laminin has been shown to play a role in cell adhesion and attachment in vivo and in vitro.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Laminin gamma3; Laminin subunit gamma-3; laminin, gamma 3; Laminin-12 subunit gamma; Laminin-14 subunit gamma; Laminin-15 subunit gamma
Gene Aliases: LAMC3; OCCM
UniProt ID: (Human) Q9Y6N6
Entrez Gene ID: (Human) 10319
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support