Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Endotoxin level < 0.01EU/µg by LAL method
Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Lanadelumab, also known as DX-2930, is a human IgG1 monoclonal antibody designed for subcutaneous self-injection. It is a fully human immunoglobulin, k-light-chain made in recombinant Chinese Hamster Ovary cells. The FDA and EU granted the designation of priority review, breakthrough therapy and orphan pharmaceutical for rare diseases based on the results of the reported clinical trials. Lanadelumab was developed by Shire and FDA approved on August 28, 2018. 5.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DX-2930; X124-G01
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support