Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ABS 022-11B-01 detects Leptin in human samples.
ABS 022-11B-01 has been successfully used in ELISA procedures. ABS 022-11B-01 binds free Leptin in solution. In sandwich ELISA, ABS 022-11B-01 works as a detection antibody with ABS 022-03-02 as capture antibody.
The ABS 022-11B-01 immunogen is a purified, recombinant Leptin protein.
NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
Leptin is a 167 amino acid long protein hormone with important effects in regulating body weight, metabolism and reproductive function. Leptin is approximately 16 kDa in mass and encoded by the obese (ob) gene. Leptin is expressed predominantly by adipocytes, which fits with the idea that body weight is sensed as the total mass of fat in the body. Smaller amounts of leptin are also secreted by cells in the epithelium of the stomach and in the placenta. Leptin receptors are highly expressed in areas of the hypothalamus known to be important in regulating body weight, as well as in T lymphocytes and vascular endothelial cells. Leptin's effects on body weight are mediated through effects on hypothalamic centers that control feeding behavior and hunger, body temperature and energy expenditure. Leptin is involved in regulating food intake, energy expenditure, and adiposity through hypothalamic leptin receptors. Leptin promotes hematopoiesis, angiogenesis, wound healing, inflammation, immune responses, influences pubertal development and fetal growth. Studies have investigated the role of leptin in obesity, anorexia nervosa, insulin resistance, and hypertension. Leptin also has thermogenic actions and regulates enzymes of fatty acid oxidation. Severe hereditary obesity in rodents and humans can be caused by defects in leptin production.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD295 antigen; H-Leptin; HuB219; LEP-R; Leptin; leptin (murine obesity homolog); leptin (obesity homolog, mouse); OB-R; Obese protein; obese, mouse, homolog of; obesity; Obesity factor; obesity protein
Gene Aliases: LEP; LEPD; OB; OBS
UniProt ID: (Human) P41159
Entrez Gene ID: (Human) 3952
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support