Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PPFIA1 may regulate the disassembly of focal adhesions. May localize receptor-like tyrosine phosphatases type 2A at specific sites on the plasma membrane, possibly regulating their interaction with the extracellular environment and their association with substrates.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ41337; FLJ42630; FLJ43474; LAR-interacting protein 1; LIP-1; liprin; Liprin-alpha-1; Liprin-alpha1; MGC26800; Protein tyrosine phosphatase receptor type f polypeptide-interacting protein alpha-1; protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 1; protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein, alpha 1; PTPRF-interacting protein alpha-1
Gene Aliases: C030014K08Rik; C87158; LIP.1; LIP1; LIPRIN; PPFIA1
UniProt ID: (Human) Q13136
Entrez Gene ID: (Human) 8500, (Rat) 293645, (Mouse) 233977
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support