Invitrogen antibodies—maximize lab budgets with advanced validation & coverage

Shop antibodies

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Aspire Free Products
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Custom Products & Projects
              • Instrument Management

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • Lirilumab Antibodies

            Invitrogen

            Lirilumab Recombinant Human Monoclonal Antibody

            View all (2) Lirilumab antibodies
            Datasheet
            Protocols
            Questions & Answers
            Tech Support
            Datasheet
            Protocols
            Questions & Answers
            Tech Support

            Cite Lirilumab Recombinant Human Monoclonal Antibody

            • Antibody Testing Data (2)
            Application
            Lirilumab Antibody in ELISA (ELISA)
            Group 53 Created with Sketch.

            FIGURE: 1 / 2

            Lirilumab Antibody (MA5-41973) in ELISA

            Direct ELISA results against the Human CD158b1/KIR2DL2 protein using a Lirilumab biosimilar monoclonal antibody (Product # MA5-41973) at a dilution of 1:5,000-1:10,000.
            View Product

            Lirilumab Antibody in ELISA (ELISA)
            Lirilumab Antibody in SDS-PAGE (SDS-PAGE)

            Product Details

            MA5-41973

            Applications
            Tested Dilution
            Publications

            ELISA (ELISA)

            1:5,000-1:10,000
            -
            Product Specifications

            Species Reactivity

            Human

            Host/Isotype

            Human / IgG4, kappa

            Expression System

            CHO cells

            Class

            Recombinant Monoclonal

            Type

            Antibody

            Immunogen

            Human KIRD2 subgroup

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            1 mg/mL

            Purification

            Protein A

            Storage buffer

            PBS, pH 7.4

            Contains

            no preservative

            Storage conditions

            -20°C, Avoid Freeze/Thaw Cycles

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2911116

            Product Specific Information

            Endotoxin level < 0.01EU/µg by LAL method

            Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.

            Target Information

            Lirilumab, also known as IPH2102 or BMS-986015, is a fully human IgG4 monoclonal antibody targeting inhibitory killer-cell immunoglobulin-like receptors (KIRs), notably KIR2DL1, KIR2DL2, and KIR2DL3, as well as activating KIR2DS1 and KIR2DS2. The structure of lirilumab includes the variable domains which bind to a conformational epitope on domain 1 of KIR2DL3. This binding does not significantly alter the structure of KIR2DL3 in comparison to its unbound form. Lirilumab's binding epitope overlaps with the HLA binding site, which influences the clinical efficacy of the drug as variations in this epitope among patients may impact its effectiveness. Functionally, lirilumab blocks inhibitory KIRs, thereby enhancing NK cell activity against tumor cells. This mechanism underlies its investigation in clinical trials for hematologic and solid malignancies, evaluating its efficacy both as a monotherapy and in combination with other agents such as azacitidine.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us About Us
            • Careers Careers
            • Investors Investors
            • News News
            • Social Responsibility Social Responsibility
            • Trademarks
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Policy
            • Price & Freight Policy
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            hong kong flag icon
            Hong Kong

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-f2c6t:80/100.66.78.227:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0