Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Loquacious (Loqs) interacts with Dicer1 (Dcr1) to facilitate miRNA processing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CG6866-PA; CG6866-PB; CG6866-PC; CG6866-PD; CG6866-PE; CG6866-PF; double-stranded RNA binding protein; Dsim/loqs; Dsimloqs; Dsimw501_GD23896; dsRNA-binding protein; loqs-PA; loqs-PB; loqs-PC; loqs-PD; loqs-PE; loqs-PF; Loquacious,isoform B; Protein Loquacious; RE14437p
Gene Aliases: CG6866; Dmel\CG6866; Dmel_CG6866; dRax; loq; LOQS; Loqs-PD; LqPD; R3D1; R3D1-L; R3D1-S; TRBP
UniProt ID: (Fruit fly) Q9VJY9
Entrez Gene ID: (Fruit fly) 34751
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support