Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • Ly-6G/Ly-6C Antibodies

            Invitrogen

            Ly-6G/Ly-6C Monoclonal Antibody (RB6-8C5), eBioscience™

            Advanced Verification
            94 Published Figures
            383 References
            View all (62) Ly-6G/Ly-6C antibodies
            Datasheet
            Protocols
            Questions & Answers
            Catalog # 14-5931-82
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (JPY)
            18,900
             
            Contact Us
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite Ly-6G/Ly-6C Monoclonal Antibody (RB6-8C5), eBioscience™

            • Antibody Testing Data (1)
            • Published Figures (94)
            • Advanced Verification (2)
            Application
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Group 53 Created with Sketch.

            FIGURE: 1 / 97

            Ly-6G/Ly-6C Antibody (14-5931-82) in Flow

            Staining of mouse bone marrow with Anti-Mouse Ly-6G (Gr-1) FITC and Anti-Human/Mouse CD45R (B220) PE (Product # 12-0452-82).Total viable cells were used for analysis.
            View Product

            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
            Ly-6G/Ly-6C Antibody in Immunocytochemistry (ICC/IF)
            Ly-6G/Ly-6C Antibody in Immunocytochemistry (ICC/IF)
            Ly-6G/Ly-6C Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
            Ly-6G/Ly-6C Antibody in Immunocytochemistry (ICC/IF)
            Ly-6G/Ly-6C Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
            Ly-6G/Ly-6C Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
            Ly-6G/Ly-6C Antibody in Immunocytochemistry (ICC/IF)
            Ly-6G/Ly-6C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Ly-6G/Ly-6C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Ly-6G/Ly-6C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Ly-6G/Ly-6C Antibody in Immunohistochemistry (Frozen) (IHC (F))
            Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
            Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
            Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
            Ly-6G/Ly-6C Antibody in Western Blot, Immunohistochemistry (WB, IHC)
            Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
            Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
            Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
            Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
            Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
            Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
            Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
            Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
            Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
            Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
            Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
            Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
            Ly-6G/Ly-6C Antibody
            Ly-6G/Ly-6C Antibody

            Product Details

            14-5931-82

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            Assay-Dependent
            View 3 publications 3 publications

            Immunohistochemistry (IHC)

            -
            View 60 publications 60 publications

            Immunohistochemistry (Paraffin) (IHC (P))

            10 µg/mL
            View 6 publications 6 publications

            Immunohistochemistry (Frozen) (IHC (F))

            10 µg/mL
            View 9 publications 9 publications

            Immunocytochemistry (ICC/IF)

            -
            View 19 publications 19 publications

            Flow Cytometry (Flow)

            0.5 µg/test
            View 238 publications 238 publications

            Immunoprecipitation (IP)

            Assay-Dependent
            View 2 publications 2 publications

            Neutralization (Neu)

            -
            View 5 publications 5 publications

            Functional Assay (Functional)

            Assay-Dependent
            View 28 publications 28 publications

            Inhibition Assays (IA)

            -
            View 1 publication 1 publication

            In Situ Hybridization (ISH)

            -
            View 1 publication 1 publication

            In vitro Assay (IV)

            -
            View 1 publication 1 publication

            Miscellaneous PubMed (Misc)

            -
            View 10 publications 10 publications
            Product Specifications

            Species Reactivity

            Mouse

            Published species

            Dog, Human, Mouse, Rat

            Host/Isotype

            Rat / IgG2b, kappa

            Class

            Monoclonal

            Type

            Antibody

            Clone

            RB6-8C5

            Conjugate

            Unconjugated
            • Alexa Fluor 488 
            • Alexa Fluor 532 
            • Alexa Fluor 561
            • Alexa Fluor 660 
            • Alexa Fluor 700 
            • APC 
            • APC-eFluor 780
            • Biotin 
            • Brilliant UV 395
            • Brilliant UV 496
            • Brilliant UV 563
            • Brilliant UV 615
            • Brilliant UV 661
            • Brilliant UV 805
            • Brilliant Violet 421
            • Brilliant Violet 650
            • Brilliant Violet 711
            • Brilliant Violet 786
            • eFluor 450
            • eFluor 506
            • eFluor 660
            • FITC
            • Functional Grade
            • NovaFluor Red 700
            • PE 
            • PE-Cyanine5
            • PE-Cyanine5.5
            • PE-Cyanine7
            • PE-eFluor 610
            • PerCP-Cyanine5.5
            • PerCP-eFluor 710
            • Super Bright 436
            • Super Bright 600
            • Super Bright 645
            • Super Bright 702
            • Super Bright 780
            • Request custom conjugation

            Form

            Liquid

            Concentration

            0.5 mg/mL

            Purification

            Affinity chromatography

            Storage buffer

            PBS, pH 7.2

            Contains

            0.09% sodium azide

            Storage conditions

            4°C

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_467730

            Product Specific Information

            Description: The RB6-8C5 monoclonal antibody reacts with mouse Ly-6G, a 21-25 kDa protein also known as the myeloid differentiation antigen Gr-1. A GPI-linked protein, Gr-1 is expressed by the myeloid lineage in a developmentally regulated manner in the bone marrow. While monocytes only express Gr-1 transiently during their bone marrow development, the expression of Gr-1 on bone marrow granulocytes as well as on peripheral neutrophils is a good marker for these populations.

            eBioscience testing indicates that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also stains cells that express the highest levels of Ly6c (as defined by staining with antibody clone HK1.4). It is recommended that 1A8-Ly6G (Product # 9668) be used when looking at Ly-6G specific targets.

            Applications Reported: The RB6-8C5 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, immunoblotting (WB), and immunohistochemical staining of frozen or formalin-fixed paraffin embedded tissue. It has also been reported in cell depletion. (Please use Functional Grade purified RB6-8C5,Product # 16-5931-82, in functional assays).

            Applications Tested: The RB6-8C5 antibody has been tested by flow cytometric analysis of mouse bone marrow cells and splenocytes. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. The RB6-8C5 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded mouse tissue using low or high pH antigen retrieval and can be used at less than or equal to 10 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

            Purity: Greater than 90%, as determined by SDS-PAGE.

            Aggregation: Less than 10%, as determined by HPLC.

            Filtration: 0.2 µm post-manufacturing filtered.

            Target Information

            Granulocytes are a category of white blood cells characterised by the presence of granules in their cytoplasm. They are also called polymorphonuclear leukocytes (PMN or PML) because of the varying shapes of the nucleus, which is usually lobed into three segments. In common parlance, the term polymorphonuclear leukocyte often refers specifically to neutrophil granulocytes, the most abundant of the granulocytes. Granulocytes or PMN are released from the bone marrow by the regulatory complement proteins.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Related Products for

            Bioinformatics

            Protein Aliases: Granulocyte Marker; Ly-6C1; Ly-6C2; Ly-6G; Ly-6G.1; lymphocyte antigen 6 complex, locus C; lymphocyte antigen 6C precursor (Ly-6C); Lymphocyte antigen 6C1; Lymphocyte antigen 6C2; Lymphocyte antigen 6G; Lymphocyte antigen Ly-6C; lymphocyte differentiation antigen; PML; PMN

            View more View less

            Gene Aliases: AA682074; AA959465; Gr-1; Gr1; Ly-6C; Ly-6C.2; Ly-6C1; Ly-6C2; Ly-6G; Ly6c; Ly6c1; Ly6c2; Ly6g

            View more View less

            UniProt ID: (Mouse) P0CW02, (Mouse) P0CW03, (Mouse) P35461

            View more View less

            Entrez Gene ID: (Mouse) 17067, (Mouse) 100041546, (Mouse) 546644

            View more View less

            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            japan | 日本 flag icon
            Japan | 日本

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-flzw4:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0