Product References
Nonredundancy of IL-1α and IL-1β is defined by distinct regulation of tissues orchestrating resistance versus tolerance to infection.
Science advances
Eislmayr K,Bestehorn A,Morelli L,Borroni M,Vande Walle L,Lamkanfi M,Kovarik P
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunohistochemistry
Fri Mar 04 00:00:00 EST 2022
Chronic T cell proliferation in brains after stroke could interfere with the efficacy of immunotherapies.
The Journal of experimental medicine
Heindl S,Ricci A,Carofiglio O,Zhou Q,Arzberger T,Lenart N,Franzmeier N,Hortobagyi T,Nelson PT,Stowe AM,Denes A,Edbauer D,Liesz A
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Mon Aug 02 00:00:00 EDT 2021
RIPK3 Facilitates Host Resistance to Oral Toxoplasma gondii Infection.
Infection and immunity
Cervantes PW,Martorelli Di Genova B,Erazo Flores BJ,Knoll LJ
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunohistochemistry
Fri Apr 16 00:00:00 EDT 2021
BRD4-mediated repression of p53 is a target for combination therapy in AML.
Nature communications
Latif AL,Newcombe A,Li S,Gilroy K,Robertson NA,Lei X,Stewart HJS,Cole J,Terradas MT,Rishi L,McGarry L,McKeeve C,Reid C,Clark W,Campos J,Kirschner K,Davis A,Lopez J,Sakamaki JI,Morton JP,Ryan KM,Tait SWG,Abraham SA,Holyoake T,Higgins B,Huang X,Blyth K,Copland M,Chevassut TJT,Keeshan K,Adams PD
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Mon Jan 11 00:00:00 EST 2021
Design, Synthesis and Biological Evaluation of Arylpyridin-2-yl Guanidine Derivatives and Cyclic Mimetics as Novel MSK1 Inhibitors. An Application in an Asthma Model.
Molecules (Basel, Switzerland)
Bollenbach M,Nemska S,Wagner P,Camelin G,Daubeuf F,Obrecht A,Villa P,Rognan D,Bihel F,Bourguignon JJ,Schmitt M,Frossard N
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Wed Jan 13 00:00:00 EST 2021
IFNγ-Producing γ/δ T Cells Accumulate in the Fetal Brain Following Intrauterine Inflammation.
Frontiers in immunology
Lewis EL,Tulina N,Anton L,Brown AG,Porrett PM,Elovitz MA
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Wed Jan 05 00:00:00 EST 2022
Lung mesenchymal stromal cells influenced by Th2 cytokines mobilize neutrophils and facilitate metastasis by producing complement C3.
Nature communications
Zheng Z,Li YN,Jia S,Zhu M,Cao L,Tao M,Jiang J,Zhan S,Chen Y,Gao PJ,Hu W,Wang Y,Shao C,Shi Y
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Wed Oct 27 00:00:00 EDT 2021
Retinoic Acid Inhibits Tumor-Associated Mesenchymal Stromal Cell Transformation in Melanoma.
Frontiers in cell and developmental biology
Lou Q,Zhao M,Xu Q,Xie S,Liang Y,Chen J,Yuan L,Wang L,Jiang L,Mou L,Lin D,Zhao M
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Sat Apr 24 00:00:00 EDT 2021
Comprehensive mutagenesis identifies the peptide repertoire of a p53 T-cell receptor mimic antibody that displays no toxicity in mice transgenic for human HLA-A*0201.
PloS one
Trenevska I,Anderson AP,Bentley C,Hassanali T,Wiblin S,Maguire S,Pezzella F,Banham AH,Li D
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Wed Sep 29 00:00:00 EDT 2021
Abraxane-induced bone marrow CD11b+ myeloid cell depletion in tumor-bearing mice is visualized by μPET-CT with 64Cu-labeled anti-CD11b and prevented by anti-CSF-1.
Theranostics
Cao Q,Huang Q,Wang YA,Li C
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Fri Jul 23 00:00:00 EDT 2021
SPARC regulation of PMN clearance protects from pristane-induced lupus and rheumatoid arthritis.
iScience
Sangaletti S,Botti L,Gulino A,Lecis D,Bassani B,Portararo P,Milani M,Cancila V,De Cecco L,Dugo M,Tripodo C,Colombo MP
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunohistochemistry
Fri Jun 25 00:00:00 EDT 2021
Single-cell analyses unravel cell type-specific responses to a vitamin D analog in prostatic precancerous lesions.
Science advances
Abu El Maaty MA,Grelet E,Keime C,Rerra AI,Gantzer J,Emprou C,Terzic J,Lutzing R,Bornert JM,Laverny G,Metzger D
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Thu Jul 01 00:00:00 EDT 2021
Construction, characterization, and immunization of nanoparticles that display a diverse array of influenza HA trimers.
PloS one
Cohen AA,Yang Z,Gnanapragasam PNP,Ou S,Dam KA,Wang H,Bjorkman PJ
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Fri Oct 15 00:00:00 EDT 2021
Calibration of cell-intrinsic interleukin-2 response thresholds guides design of a regulatory T cell biased agonist.
eLife
Glassman CR,Su L,Majri-Morrison SS,Winkelmann H,Mo F,Li P,Pérez-Cruz M,Ho PP,Koliesnik I,Nagy N,Hnizdilova T,Picton LK,Kovar M,Bollyky P,Steinman L,Meyer E,Piehler J,Leonard WJ,Garcia KC
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Tue May 18 00:00:00 EDT 2021
Colonization of dermal arterioles by Neisseria meningitidis provides a safe haven from neutrophils.
Nature communications
Manriquez V,Nivoit P,Urbina T,Echenique-Rivera H,Melican K,Fernandez-Gerlinger MP,Flamant P,Schmitt T,Bruneval P,Obino D,Duménil G
16-5931-82 was used in Flow Cytometry to demonstrate how neutrophil recruitment contributes to resolution of Neisseria meningitidis in a humanized mouse model.
Tue Jul 27 00:00:00 EDT 2021
Deregulated Notch and Wnt signaling activates early-stage myeloid regeneration pathways in leukemia.
The Journal of experimental medicine
Kang YA,Pietras EM,Passegué E
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Mon Mar 02 00:00:00 EST 2020
In Situ Therapeutic Cancer Vaccination with an Oncolytic Virus Expressing Membrane-Tethered IL-2.
Molecular therapy oncolytics
Liu W,Dai E,Liu Z,Ma C,Guo ZS,Bartlett DL
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Fri Jun 26 00:00:00 EDT 2020
Deficiency of immunoglobulin E protects mice from experimental abdominal aortic aneurysms.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Li J,Deng Z,Zhang X,Liu F,Yang C,Shi GP
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Sat Feb 01 00:00:00 EST 2020
Flagellin-Stimulated Production of Interferon-β Promotes Anti-Flagellin IgG2c and IgA Responses.
Molecules and cells
Kang W,Park A,Huh JW,You G,Jung DJ,Song M,Lee HK,Kim YM
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Tue Mar 31 00:00:00 EDT 2020
Effects of Hypertonic Saline and Hydroxyethyl Starch on Myeloid-Derived Suppressor Cells in Hemorrhagic Shock Mice under Secondary Bacterial Attack.
BioMed research international
Jiang JK,Hong LJ,Lu YQ
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Wed Dec 16 00:00:00 EST 2020
Long Non-Coding RNA Hotairm1 Promotes S100A9 Support of MDSC Expansion during Sepsis.
Journal of clinical & cellular immunology
Alkhateeb T,Bah I,Kumbhare A,Youssef D,Yao ZQ,McCall CE,Gazzar ME
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Sat Aug 13 00:00:00 EDT 2022
The lysophospholipase D enzyme Gdpd3 is required to maintain chronic myelogenous leukaemia stem cells.
Nature communications
Naka K,Ochiai R,Matsubara E,Kondo C,Yang KM,Hoshii T,Araki M,Araki K,Sotomaru Y,Sasaki K,Mitani K,Kim DW,Ooshima A,Kim SJ
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Thu Sep 17 00:00:00 EDT 2020
Hyaluronan Degradation by Cemip Regulates Host Defense against Staphylococcus aureus Skin Infection.
Cell reports
Dokoshi T,Zhang LJ,Li F,Nakatsuji T,Butcher A,Yoshida H,Shimoda M,Okada Y,Gallo RL
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Tue Jan 07 00:00:00 EST 2020
Renal proximal tubular NEMO plays a critical role in ischemic acute kidney injury.
JCI insight
Han SJ,Williams RM,Kim M,Heller DA,D'Agati V,Schmidt-Supprian M,Lee HT
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunohistochemistry
Thu Sep 17 00:00:00 EDT 2020
Loss of testosterone impairs anti-tumor neutrophil function.
Nature communications
Markman JL,Porritt RA,Wakita D,Lane ME,Martinon D,Noval Rivas M,Luu M,Posadas EM,Crother TR,Arditi M
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Tue Mar 31 00:00:00 EDT 2020
Bacterial and Fungal Toll-Like Receptor Activation Elicits Type I IFN Responses in Mast Cells.
Frontiers in immunology
Kornstädt L,Pierre S,Weigert A,Ebersberger S,Schäufele TJ,Kolbinger A,Schmid T,Cohnen J,Thomas D,Ferreirós N,Brüne B,Ebersberger I,Scholich K
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Mon Jun 21 00:00:00 EDT 2021
Sensing and signaling of immunogenic extracellular RNAs restrain group 2 innate lymphoid cell-driven acute lung inflammation and airway hyperresponsiveness.
PloS one
She L,Alanazi HH,Yan L,Brooks EG,Dube PH,Xiang Y,Zhang F,Sun Y,Liu Y,Zhang X,Li XD
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Thu Sep 24 00:00:00 EDT 2020
Prevention of acute kidney injury by low intensity pulsed ultrasound via anti-inflammation and anti-apoptosis.
Scientific reports
Chiang CK,Loh JZ,Yang TH,Huang KT,Wu CT,Guan SS,Liu SH,Hung KY
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunohistochemistry
Mon Aug 31 00:00:00 EDT 2020
Prevention of acute kidney injury by low intensity pulsed ultrasound via anti-inflammation and anti-apoptosis.
Scientific reports
Chiang CK,Loh JZ,Yang TH,Huang KT,Wu CT,Guan SS,Liu SH,Hung KY
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunohistochemistry
Mon Aug 31 00:00:00 EDT 2020
Protocol for Flow Cytometric Detection of Immune Cell Infiltration in the Epidermis and Dermis of a Psoriasis Mouse Model.
STAR protocols
Lou F,Sun Y,Wang H
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Fri Dec 18 00:00:00 EST 2020
The endothelial protein C receptor plays an essential role in the maintenance of pregnancy.
Science advances
Castillo MM,Yang Q,Sigala AS,McKinney DT,Zhan M,Chen KL,Jarzembowski JA,Sood R
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunohistochemistry
Sun Nov 01 00:00:00 EDT 2020
Neutrophils contribute to spontaneous resolution of liver inflammation and fibrosis via microRNA-223.
The Journal of clinical investigation
Calvente CJ,Tameda M,Johnson CD,Del Pilar H,Lin YC,Adronikou N,De Mollerat Du Jeu X,Llorente C,Boyer J,Feldstein AE
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Thu Jul 11 00:00:00 EDT 2019
The role of platelets in mediating a response to human influenza infection.
Nature communications
Koupenova M,Corkrey HA,Vitseva O,Manni G,Pang CJ,Clancy L,Yao C,Rade J,Levy D,Wang JP,Finberg RW,Kurt-Jones EA,Freedman JE
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunocytochemistry
Tue Apr 16 00:00:00 EDT 2019
Neutrophil extracellular traps induced by VP1 contribute to pulmonary edema during EV71 infection.
Cell death discovery
Wang N,Yang X,Sun J,Sun Z,Ma Q,Wang Z,Chen Z,Wang Z,Hu F,Wang H,Zhou L,Zhang M,Xu J
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Wed Sep 30 00:00:00 EDT 2020
DNA released from neutrophil extracellular traps (NETs) activates pancreatic stellate cells and enhances pancreatic tumor growth.
Oncoimmunology
Miller-Ocuin JL,Liang X,Boone BA,Doerfler WR,Singhi AD,Tang D,Kang R,Lotze MT,Zeh HJ
16-5931-82 was used in Immunohistochemistry to support further investigation into targeting of NETs, PADI4 and extracellular DNA as a potential treatment strategy in patients with pancreatic cancer.
Wed Feb 03 00:00:00 EST 2021
E3 Ligase VHL Promotes Group 2 Innate Lymphoid Cell Maturation and Function via Glycolysis Inhibition and Induction of Interleukin-33 Receptor.
Immunity
Li Q,Li D,Zhang X,Wan Q,Zhang W,Zheng M,Zou L,Elly C,Lee JH,Liu YC
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Tue Feb 20 00:00:00 EST 2018
Inactivation of mTORC1 Signaling in Osterix-Expressing Cells Impairs B-cell Differentiation.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research
Wang Y,Xiao M,Tao C,Chen J,Wang Z,Yang J,Chen Z,Zou Z,Liu A,Cai D,Jiang Y,Ding C,Li M,Bai X
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Sun Apr 01 00:00:00 EDT 2018
Frontline Science: Macrophage-derived exosomes promote neutrophil necroptosis following hemorrhagic shock.
Journal of leukocyte biology
Jiao Y,Li Z,Loughran PA,Fan EK,Scott MJ,Li Y,Billiar TR,Wilson MA,Shi X,Fan J
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Thu Feb 01 00:00:00 EST 2018
Dual specificity phosphatase DUSP6 promotes endothelial inflammation through inducible expression of ICAM-1.
The FEBS journal
Hsu SF,Lee YB,Lee YC,Chung AL,Apaya MK,Shyur LF,Cheng CF,Ho FM,Meng TC
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Tue May 01 00:00:00 EDT 2018
Investigation of tumor-tumor interactions in a double human cervical carcinoma xenograft model in nude mice.
Oncotarget
Mertens B,Cristina de Araujo Nogueira T,Topalis D,Stranska R,Snoeck R,Andrei G
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Tue Apr 24 00:00:00 EDT 2018
Analysis of leukocyte transepithelial migration using an in vivo murine colonic loop model.
JCI insight
Flemming S,Luissint AC,Nusrat A,Parkos CA
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunofluorescence
Thu Oct 18 00:00:00 EDT 2018
Antiviral effects of ferric ammonium citrate.
Cell discovery
Wang H,Li Z,Niu J,Xu Y,Ma L,Lu A,Wang X,Qian Z,Huang Z,Jin X,Leng Q,Wang J,Zhong J,Sun B,Meng G
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Tue Sep 29 00:00:00 EDT 2020
Induction of neutrophil apoptosis by a Bcl-2 inhibitor reduces particulate matter-induced lung inflammation.
Aging
Geng X,Wang X,Luo M,Xing M,Wu Y,Li W,Chen Z,Shen H,Ying S
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Tue Jun 26 00:00:00 EDT 2018
Microbiota Reconstitution Does Not Cause Bone Loss in Germ-Free Mice.
mSphere
Quach D,Collins F,Parameswaran N,McCabe L,Britton RA
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Wed Nov 20 00:00:00 EST 2019
Epithelial disruptions, but not immune cell invasion, induced secretory dysfunction following innate immune activation in a novel model of acute salivary gland injury.
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology
Shaalan A,Carpenter G,Proctor G
16-5931-82 was used in In vivo experiments to develop a novel injury model of the SGs, featuring acute secretory dysfunction and immediate structural disruptions, ruling out injurious role of aggressively infiltrating inflammatory cells.
Thu Feb 01 00:00:00 EST 2018
Colony stimulating factor-1 receptor is a central component of the foreign body response to biomaterial implants in rodents and non-human primates.
Nature materials
Doloff JC,Veiseh O,Vegas AJ,Tam HH,Farah S,Ma M,Li J,Bader A,Chiu A,Sadraei A,Aresta-Dasilva S,Griffin M,Jhunjhunwala S,Webber M,Siebert S,Tang K,Chen M,Langan E,Dholokia N,Thakrar R,Qi M,Oberholzer J,Greiner DL,Langer R,Anderson DG
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Thu Jun 01 00:00:00 EDT 2017
Brd4 modulates the innate immune response through Mnk2-eIF4E pathway-dependent translational control of IκBα.
Proceedings of the National Academy of Sciences of the United States of America
Bao Y,Wu X,Chen J,Hu X,Zeng F,Cheng J,Jin H,Lin X,Chen LF
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Tue May 16 00:00:00 EDT 2017
Cardiac inflammation in genetic dilated cardiomyopathy caused by MYBPC3 mutation.
Journal of molecular and cellular cardiology
Lynch TL,Ismahil MA,Jegga AG,Zilliox MJ,Troidl C,Prabhu SD,Sadayappan S
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunocytochemistry
Sun Jan 01 00:00:00 EST 2017
Silybin attenuates LPS-induced lung injury in mice by inhibiting NF-κB signaling and NLRP3 activation.
International journal of molecular medicine
Zhang B,Wang B,Cao S,Wang Y,Wu D
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Mon May 01 00:00:00 EDT 2017
ADAR1 deletion induces NFκB and interferon signaling dependent liver inflammation and fibrosis.
RNA biology
Ben-Shoshan SO,Kagan P,Sultan M,Barabash Z,Dor C,Jacob-Hirsch J,Harmelin A,Pappo O,Marcu-Malina V,Ben-Ari Z,Amariglio N,Rechavi G,Goldstein I,Safran M
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Thu May 04 00:00:00 EDT 2017
Coinjection of IL2 DNA enhances E7-specific antitumor immunity elicited by intravaginal therapeutic HPV DNA vaccination with electroporation.
Gene therapy
Sun Y,Peng S,Yang A,Farmer E,Wu TC,Hung CF
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Sat Jul 01 00:00:00 EDT 2017
MiR-125a Is a critical modulator for neutrophil development.
PLoS genetics
Qin Y,Wu L,Ouyang Y,Zhou P,Zhou H,Wang Y,Ma J,Zhang J,Chen Y,Qian J,Tang Y,Shen N
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Sun Oct 01 00:00:00 EDT 2017
Gastric TFF1 Expression from Acute to Chronic Helicobacter Infection.
Frontiers in cellular and infection microbiology
Esposito R,Morello S,Vllahu M,Eletto D,Porta A,Tosco A
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunohistochemistry
Mon Jul 09 00:00:00 EDT 2018
Recipient bone marrow assimilates the myeloid/lymphoid reconstitution of distinct fetal hematopoietic stem cells.
Oncotarget
Guo XL,Chu L,Ke F,Mu LL,Li Z,Cai JJ,Li HF,Hong DL
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Tue Dec 12 00:00:00 EST 2017
BOT-4-one attenuates NLRP3 inflammasome activation: NLRP3 alkylation leading to the regulation of its ATPase activity and ubiquitination.
Scientific reports
Shim DW,Shin WY,Yu SH,Kim BH,Ye SK,Koppula S,Won HS,Kang TB,Lee KH
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Wed Nov 08 00:00:00 EST 2017
Tumor-Derived Exosomes Induce the Formation of Neutrophil Extracellular Traps: Implications For The Establishment of Cancer-Associated Thrombosis.
Scientific reports
Leal AC,Mizurini DM,Gomes T,Rochael NC,Saraiva EM,Dias MS,Werneck CC,Sielski MS,Vicente CP,Monteiro RQ
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunohistochemistry
Tue Jul 25 00:00:00 EDT 2017
One Year Follow-Up Risk Assessment in SKH-1 Mice and Wounds Treated with an Argon Plasma Jet.
International journal of molecular sciences
Schmidt A,Woedtke TV,Stenzel J,Lindner T,Polei S,Vollmar B,Bekeschus S
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunofluorescence
Wed Apr 19 00:00:00 EDT 2017
Combinatorial hydrogel library enables identification of materials that mitigate the foreign body response in primates.
Nature biotechnology
Vegas AJ,Veiseh O,Doloff JC,Ma M,Tam HH,Bratlie K,Li J,Bader AR,Langan E,Olejnik K,Fenton P,Kang JW,Hollister-Locke J,Bochenek MA,Chiu A,Siebert S,Tang K,Jhunjhunwala S,Aresta-Dasilva S,Dholakia N,Thakrar R,Vietti T,Chen M,Cohen J,Siniakowicz K,Qi M,McGarrigle J,Graham AC,Lyle S,Harlan DM,Greiner DL,Oberholzer J,Weir GC,Langer R,Anderson DG
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Tue Mar 01 00:00:00 EST 2016
The Deubiquitinase OTULIN Is an Essential Negative Regulator of Inflammation and Autoimmunity.
Cell
Damgaard RB,Walker JA,Marco-Casanova P,Morgan NV,Titheradge HL,Elliott PR,McHale D,Maher ER,McKenzie ANJ,Komander D
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Thu Aug 25 00:00:00 EDT 2016
Multiple myeloma exosomes establish a favourable bone marrow microenvironment with enhanced angiogenesis and immunosuppression.
The Journal of pathology
Wang J,De Veirman K,Faict S,Frassanito MA,Ribatti D,Vacca A,Menu E
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Wed Jun 01 00:00:00 EDT 2016
Opposite Effects of Coinjection and Distant Injection of Mesenchymal Stem Cells on Breast Tumor Cell Growth.
Stem cells translational medicine
Zheng H,Zou W,Shen J,Xu L,Wang S,Fu YX,Fan W
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Thu Sep 01 00:00:00 EDT 2016
Macrophages transfer antigens to dendritic cells by releasing exosomes containing dead-cell-associated antigens partially through a ceramide-dependent pathway to enhance CD4(+) T-cell responses.
Immunology
Xu Y,Liu Y,Yang C,Kang L,Wang M,Hu J,He H,Song W,Tang H
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Sat Oct 01 00:00:00 EDT 2016
Effects of ketanserin on experimental colitis in mice and macrophage function.
International journal of molecular medicine
Xiao J,Shao L,Shen J,Jiang W,Feng Y,Zheng P,Liu F
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Tue Mar 01 00:00:00 EST 2016
The miR-17∼92 cluster contributes to MLL leukemia through the repression of MEIS1 competitor PKNOX1.
Leukemia research
Mian YA,Zeleznik-Le NJ
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Fri Jul 01 00:00:00 EDT 2016
Murine iPSC-Derived Macrophages as a Tool for Disease Modeling of Hereditary Pulmonary Alveolar Proteinosis due to Csf2rb Deficiency.
Stem cell reports
Mucci A,Kunkiel J,Suzuki T,Brennig S,Glage S,Kühnel MP,Ackermann M,Happle C,Kuhn A,Schambach A,Trapnell BC,Hansen G,Moritz T,Lachmann N
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Tue Aug 09 00:00:00 EDT 2016
Activated c-Kit receptor in the heart promotes cardiac repair and regeneration after injury.
Cell death & disease
Di Siena S,Gimmelli R,Nori SL,Barbagallo F,Campolo F,Dolci S,Rossi P,Venneri MA,Giannetta E,Gianfrilli D,Feigenbaum L,Lenzi A,Naro F,Cianflone E,Mancuso T,Torella D,Isidori AM,Pellegrini M
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunofluorescence
Thu Jul 28 00:00:00 EDT 2016
Activated c-Kit receptor in the heart promotes cardiac repair and regeneration after injury.
Cell death & disease
Di Siena S,Gimmelli R,Nori SL,Barbagallo F,Campolo F,Dolci S,Rossi P,Venneri MA,Giannetta E,Gianfrilli D,Feigenbaum L,Lenzi A,Naro F,Cianflone E,Mancuso T,Torella D,Isidori AM,Pellegrini M
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunofluorescence
Thu Jul 28 00:00:00 EDT 2016
Influenza H3N2 infection of the collaborative cross founder strains reveals highly divergent host responses and identifies a unique phenotype in CAST/EiJ mice.
BMC genomics
Leist SR,Pilzner C,van den Brand JM,Dengler L,Geffers R,Kuiken T,Balling R,Kollmus H,Schughart K
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Sat Feb 27 00:00:00 EST 2016
Stroke Damage Is Exacerbated by Nano-Size Particulate Matter in a Mouse Model.
PloS one
Liu Q,Babadjouni R,Radwanski R,Cheng H,Patel A,Hodis DM,He S,Baumbacher P,Russin JJ,Morgan TE,Sioutas C,Finch CE,Mack WJ
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunofluorescence
Tue Aug 23 00:00:00 EDT 2016
C/EBP homologous protein (CHOP) deficiency ameliorates renal fibrosis in unilateral ureteral obstructive kidney disease.
Oncotarget
Liu SH,Wu CT,Huang KH,Wang CC,Guan SS,Chen LP,Chiang CK
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunohistochemistry
Tue Apr 19 00:00:00 EDT 2016
C/EBP homologous protein (CHOP) deficiency ameliorates renal fibrosis in unilateral ureteral obstructive kidney disease.
Oncotarget
Liu SH,Wu CT,Huang KH,Wang CC,Guan SS,Chen LP,Chiang CK
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunohistochemistry
Tue Apr 19 00:00:00 EDT 2016
Functional-genetic dissection of HDAC dependencies in mouse lymphoid and myeloid malignancies.
Blood
Matthews GM,Mehdipour P,Cluse LA,Falkenberg KJ,Wang E,Roth M,Santoro F,Vidacs E,Stanley K,House CM,Rusche JR,Vakoc CR,Zuber J,Minucci S,Johnstone RW
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Thu Nov 19 00:00:00 EST 2015
An anti-silencer- and SATB1-dependent chromatin hub regulates Rag1 and Rag2 gene expression during thymocyte development.
The Journal of experimental medicine
Hao B,Naik AK,Watanabe A,Tanaka H,Chen L,Richards HW,Kondo M,Taniuchi I,Kohwi Y,Kohwi-Shigematsu T,Krangel MS
16-5931 was used in Flow cytometry/Cell sorting to provide a novel framework for understanding ASE function for gene expression in double-positive thymocytes.
Mon May 04 00:00:00 EDT 2015
Redox Regulation of Pro-IL-1β Processing May Contribute to the Increased Severity of Serum-Induced Arthritis in NOX2-Deficient Mice.
Antioxidants & redox signaling
Huang YF,Lo PC,Yen CL,Nigrovic PA,Chao WC,Wang WZ,Hsu GC,Tsai YS,Shieh CC
16-5931 was used in Neutralization experiments to investigate how ROS act as a negative feedback to constrain IL-1β-mediated inflammation, accounting for the more severe arthritis in the absence of NOX2.
Tue Oct 20 00:00:00 EDT 2015
The receptor for advanced glycation end products (RAGE) enhances autophagy and neutrophil extracellular traps in pancreatic cancer.
Cancer gene therapy
Boone BA,Orlichenko L,Schapiro NE,Loughran P,Gianfrate GC,Ellis JT,Singhi AD,Kang R,Tang D,Lotze MT,Zeh HJ
16-5931 was used in Immunodepletion to demonstrate that neutrophil extracellular traps are upregulated in pancreatic cancer through RAGE dependent/autophagy pathways.
Mon Jun 01 00:00:00 EDT 2015
The receptor for advanced glycation end products (RAGE) enhances autophagy and neutrophil extracellular traps in pancreatic cancer.
Cancer gene therapy
Boone BA,Orlichenko L,Schapiro NE,Loughran P,Gianfrate GC,Ellis JT,Singhi AD,Kang R,Tang D,Lotze MT,Zeh HJ
16-5931 was used in Immunodepletion to demonstrate that neutrophil extracellular traps are upregulated in pancreatic cancer through RAGE dependent/autophagy pathways.
Mon Jun 01 00:00:00 EDT 2015
A far-red fluorescent probe for flow cytometry and image-based functional studies of xenobiotic sequestering macrophages.
Cytometry. Part A : the journal of the International Society for Analytical Cytology
Keswani RK,Yoon GS,Sud S,Stringer KA,Rosania GR
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Tue Sep 01 00:00:00 EDT 2015
Local delivery of VEGF and SDF enhances endothelial progenitor cell recruitment and resultant recovery from ischemia.
Tissue engineering. Part A
Anderson EM,Kwee BJ,Lewin SA,Raimondo T,Mehta M,Mooney DJ
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Wed Apr 01 00:00:00 EDT 2015
Reducing progression of experimental lupus nephritis via inhibition of the B7/CD28 signaling pathway.
Molecular medicine reports
Huang L,Kong Y,Wang J,Sun J,Shi Q,Qiu YH
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Tue Sep 01 00:00:00 EDT 2015
The bone marrow microenvironment enhances multiple myeloma progression by exosome-mediated activation of myeloid-derived suppressor cells.
Oncotarget
Wang J,De Veirman K,De Beule N,Maes K,De Bruyne E,Van Valckenborgh E,Vanderkerken K,Menu E
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Tue Dec 22 00:00:00 EST 2015
The HIF-1/glial TIM-3 axis controls inflammation-associated brain damage under hypoxia.
Nature communications
Koh HS,Chang CY,Jeon SB,Yoon HJ,Ahn YH,Kim HS,Kim IH,Jeon SH,Johnson RS,Park EJ
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunofluorescence
Fri Mar 20 00:00:00 EDT 2015
Glioma-associated microglia/macrophages display an expression profile different from M1 and M2 polarization and highly express Gpnmb and Spp1.
PloS one
Szulzewsky F,Pelz A,Feng X,Synowitz M,Markovic D,Langmann T,Holtman IR,Wang X,Eggen BJ,Boddeke HW,Hambardzumyan D,Wolf SA,Kettenmann H
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Tue Jan 12 00:00:00 EST 2016
Targeted Gene Correction in Osteopetrotic-Induced Pluripotent Stem Cells for the Generation of Functional Osteoclasts.
Stem cell reports
Neri T,Muggeo S,Paulis M,Caldana ME,Crisafulli L,Strina D,Focarelli ML,Faggioli F,Recordati C,Scaramuzza S,Scanziani E,Mantero S,Buracchi C,Sobacchi C,Lombardo A,Naldini L,Vezzoni P,Villa A,Ficara F
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Tue Oct 13 00:00:00 EDT 2015
Molecular mechanisms of NET formation and degradation revealed by intravital imaging in the liver vasculature.
Nature communications
Kolaczkowska E,Jenne CN,Surewaard BG,Thanabalasuriar A,Lee WY,Sanz MJ,Mowen K,Opdenakker G,Kubes P
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunofluorescence
Thu Mar 26 00:00:00 EDT 2015
Sunitinib depletes myeloid-derived suppressor cells and synergizes with a cancer vaccine to enhance antigen-specific immune responses and tumor eradication.
Oncoimmunology
Draghiciu O,Nijman HW,Hoogeboom BN,Meijerhof T,Daemen T
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Sun Mar 01 00:00:00 EST 2015
Host adaptation of a bacterial toxin from the human pathogen Salmonella Typhi.
Cell
Deng L,Song J,Gao X,Wang J,Yu H,Chen X,Varki N,Naito-Matsui Y,Galán JE,Varki A
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Thu Dec 04 00:00:00 EST 2014
Macrophages contribute to the cyclic activation of adult hair follicle stem cells.
PLoS biology
Castellana D,Paus R,Perez-Moreno M
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Mon Dec 01 00:00:00 EST 2014
Epithelial β1 integrin is required for lung branching morphogenesis and alveolarization.
Development (Cambridge, England)
Plosa EJ,Young LR,Gulleman PM,Polosukhin VV,Zaynagetdinov R,Benjamin JT,Im AM,van der Meer R,Gleaves LA,Bulus N,Han W,Prince LS,Blackwell TS,Zent R
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunofluorescence
Mon Dec 01 00:00:00 EST 2014
Bleomycin delivery by osmotic minipump: similarity to human scleroderma interstitial lung disease.
American journal of physiology. Lung cellular and molecular physiology
Lee R,Reese C,Bonner M,Tourkina E,Hajdu Z,Riemer EC,Silver RM,Visconti RP,Hoffman S
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunofluorescence
Tue Apr 15 00:00:00 EDT 2014
NADPH oxidase promotes neutrophil extracellular trap formation in pulmonary aspergillosis.
Infection and immunity
Röhm M,Grimm MJ,D'Auria AC,Almyroudis NG,Segal BH,Urban CF
16-5931 was used in Functional assays to show that NADPH oxidase is required for pulmonary clearance of Aspergillus hyphae and generation of neutrophil extracellular traps in vivo.
Thu May 01 00:00:00 EDT 2014
Anti-tumor innate immunity activated by intermittent metronomic cyclophosphamide treatment of 9L brain tumor xenografts is preserved by anti-angiogenic drugs that spare VEGF receptor 2.
Molecular cancer
Doloff JC,Chen CS,Waxman DJ
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Thu Jun 26 00:00:00 EDT 2014
Activation of NLRP3 inflammasome by crystalline structures via cell surface contact.
Scientific reports
Hari A,Zhang Y,Tu Z,Detampel P,Stenner M,Ganguly A,Shi Y
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Tue Dec 02 00:00:00 EST 2014
Selective regulation of lymphopoiesis and leukemogenesis by individual zinc fingers of Ikaros.
Nature immunology
Schjerven H,McLaughlin J,Arenzana TL,Frietze S,Cheng D,Wadsworth SE,Lawson GW,Bensinger SJ,Farnham PJ,Witte ON,Smale ST
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Tue Oct 01 00:00:00 EDT 2013
A mutual activation loop between breast cancer cells and myeloid-derived suppressor cells facilitates spontaneous metastasis through IL-6 trans-signaling in a murine model.
Breast cancer research : BCR
Oh K,Lee OY,Shon SY,Nam O,Ryu PM,Seo MW,Lee DS
16-5931-82 was used in Flow Cytometry to elucidate a novel mechanism by which myeloid-derived suppressor cells can mediate spontaneous, distant metastasis of breast cancer cells.
Thu Sep 11 00:00:00 EDT 2014
Arrayed lentiviral barcoding for quantification analysis of hematopoietic dynamics.
Stem cells (Dayton, Ohio)
Grosselin J,Sii-Felice K,Payen E,Chretien S,Tronik-Le Roux D,Leboulch P
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Tue Oct 01 00:00:00 EDT 2013
The cardioprotective protein apolipoprotein A1 promotes potent anti-tumorigenic effects.
The Journal of biological chemistry
Zamanian-Daryoush M,Lindner D,Tallant TC,Wang Z,Buffa J,Klipfell E,Parker Y,Hatala D,Parsons-Wingerter P,Rayman P,Yusufishaq MSS,Fisher EA,Smith JD,Finke J,DiDonato JA,Hazen SL
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Fri Jul 19 00:00:00 EDT 2013
R-spondin 2 signalling mediates susceptibility to fatal infectious diarrhoea.
Nature communications
Papapietro O,Teatero S,Thanabalasuriar A,Yuki KE,Diez E,Zhu L,Kang E,Dhillon S,Muise AM,Durocher Y,Marcinkiewicz MM,Malo D,Gruenheid S
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in In Situ Hybridization (ISH)
Mon Dec 30 00:00:00 EST 2013
Roles of Gr-1+ leukocytes in postincisional nociceptive sensitization and inflammation.
Anesthesiology
Sahbaie P,Li X,Shi X,Clark JD
16-5931 was used in Functional assays to identify the contribution of infiltrating neutrophils to incisional pain and inflammation.
Sat Sep 01 00:00:00 EDT 2012
Neutrophil-mediated inhibition of proinflammatory cytokine responses.
Journal of immunology (Baltimore, Md. : 1950)
Gresnigt MS,Joosten LA,Verschueren I,van der Meer JW,Netea MG,Dinarello CA,van de Veerdonk FL
16-5931 was used in Functional assays to show that polymorphonuclear neutrophils can dampen the proinflammatory response to C. albicans by protease-mediated degradation of cytokines.
Thu Nov 15 00:00:00 EST 2012
Differential gene expression in mice infected with distinct Toxoplasma strains.
Infection and immunity
Hill RD,Gouffon JS,Saxton AM,Su C
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Thu Mar 01 00:00:00 EST 2012
A model of GAG/MIP-2/CXCR2 interfaces and its functional effects.
Biochemistry
Rajasekaran D,Keeler C,Syed MA,Jones MC,Harrison JK,Wu D,Bhandari V,Hodsdon ME,Lolis EJ
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Tue Jul 17 00:00:00 EDT 2012
Preclinical evaluation of oncolytic vaccinia virus for therapy of canine soft tissue sarcoma.
PloS one
Gentschev I,Adelfinger M,Josupeit R,Rudolph S,Ehrig K,Donat U,Weibel S,Chen NG,Yu YA,Zhang Q,Heisig M,Thamm D,Stritzker J,Macneill A,Szalay AA
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunofluorescence
Thu Sep 20 00:00:00 EDT 2012
Virotherapy of canine tumors with oncolytic vaccinia virus GLV-1h109 expressing an anti-VEGF single-chain antibody.
PloS one
Patil SS,Gentschev I,Adelfinger M,Donat U,Hess M,Weibel S,Nolte I,Frentzen A,Szalay AA
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Thu Mar 28 00:00:00 EDT 2013
Virotherapy of canine tumors with oncolytic vaccinia virus GLV-1h109 expressing an anti-VEGF single-chain antibody.
PloS one
Patil SS,Gentschev I,Adelfinger M,Donat U,Hess M,Weibel S,Nolte I,Frentzen A,Szalay AA
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Thu Mar 28 00:00:00 EDT 2013
Hepatocyte-specific S100a8 and S100a9 transgene expression in mice causes Cxcl1 induction and systemic neutrophil enrichment.
Cell communication and signaling : CCS
Wiechert L,Németh J,Pusterla T,Bauer C,De Ponti A,Manthey S,Marhenke S,Vogel A,Klingmüller U,Hess J,Angel P
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Sat Dec 15 00:00:00 EST 2012
Hepatocyte-specific S100a8 and S100a9 transgene expression in mice causes Cxcl1 induction and systemic neutrophil enrichment.
Cell communication and signaling : CCS
Wiechert L,Németh J,Pusterla T,Bauer C,De Ponti A,Manthey S,Marhenke S,Vogel A,Klingmüller U,Hess J,Angel P
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Sat Dec 15 00:00:00 EST 2012
In vivo inhibition of c-MYC in myeloid cells impairs tumor-associated macrophage maturation and pro-tumoral activities.
PloS one
Pello OM,Chèvre R,Laoui D,De Juan A,Lolo F,Andrés-Manzano MJ,Serrano M,Van Ginderachter JA,Andrés V
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Mon Feb 25 00:00:00 EST 2013
Genetic deletion of chemokine receptor Ccr6 decreases atherogenesis in ApoE-deficient mice.
Circulation research
Wan W,Lim JK,Lionakis MS,Rivollier A,McDermott DH,Kelsall BL,Farber JM,Murphy PM
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Fri Aug 05 00:00:00 EDT 2011
Chemokines control naive CD8+ T cell selection of optimal lymph node antigen presenting cells.
The Journal of experimental medicine
Hickman HD,Li L,Reynoso GV,Rubin EJ,Skon CN,Mays JW,Gibbs J,Schwartz O,Bennink JR,Yewdell JW
16-5931 was used in Flow cytometry/Cell sorting to demonstrate the ability of chemokines to assist antiviral CD8(+) T cells in distinguishing between antigen presenting cells.
Mon Nov 21 00:00:00 EST 2011
Chemokines control naive CD8+ T cell selection of optimal lymph node antigen presenting cells.
The Journal of experimental medicine
Hickman HD,Li L,Reynoso GV,Rubin EJ,Skon CN,Mays JW,Gibbs J,Schwartz O,Bennink JR,Yewdell JW
16-5931 was used in Flow cytometry/Cell sorting to demonstrate the ability of chemokines to assist antiviral CD8(+) T cells in distinguishing between antigen presenting cells.
Mon Nov 21 00:00:00 EST 2011
A tritherapy combination of a fusion protein vaccine with immune-modulating doses of sequential chemotherapies in an optimized regimen completely eradicates large tumors in mice.
International journal of cancer
Song X,Guo W,Cui J,Qian X,Yi L,Chang M,Cai Q,Zhao Q
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Tue Mar 01 00:00:00 EST 2011
Brain infiltration of leukocytes contributes to the pathophysiology of temporal lobe epilepsy.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Zattoni M,Mura ML,Deprez F,Schwendener RA,Engelhardt B,Frei K,Fritschy JM
16-5931 was used in Functional assays to demonstrate that action of adaptive and innate immunity triggered locally by intrahippocampal kainate injection contributes seizure-suppressant and neuroprotective effects.
Wed Mar 16 00:00:00 EDT 2011
MMP9 is protective against lethal inflammatory mass lesions in the mouse colon.
Disease models & mechanisms
Hald A,Rønø B,Melander MC,Ding M,Holck S,Lund LR
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunohistochemistry
Tue Mar 01 00:00:00 EST 2011
DNA alkylating therapy induces tumor regression through an HMGB1-mediated activation of innate immunity.
Journal of immunology (Baltimore, Md. : 1950)
Guerriero JL,Ditsworth D,Catanzaro JM,Sabino G,Furie MB,Kew RR,Crawford HC,Zong WX
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Tue Mar 15 00:00:00 EDT 2011
Hemorrhagic shock activation of NLRP3 inflammasome in lung endothelial cells.
Journal of immunology (Baltimore, Md. : 1950)
Xiang M,Shi X,Li Y,Xu J,Yin L,Xiao G,Scott MJ,Billiar TR,Wilson MA,Fan J
16-5931 was used in Functional assays to explore a novel mechanism underlying hemorrhagic shock activation of the endothelial cell inflammasome and potential therapeutic targets for SIRS and ALI.
Tue Nov 01 00:00:00 EDT 2011
Mechanism of neutrophil recruitment to the lung after pulmonary contusion.
Shock (Augusta, Ga.)
Hoth JJ,Wells JD,Hiltbold EM,McCall CE,Yoza BK
16-5931 was used in Blocking experiments to investigate whether the innate immune and inflammatory activation of neutrophils figures prominently in the response to lung injury.
Wed Jun 01 00:00:00 EDT 2011
Brain dendritic cells in ischemic stroke: time course, activation state, and origin.
Brain, behavior, and immunity
Felger JC,Abe T,Kaunzner UW,Gottfried-Blackmore A,Gal-Toth J,McEwen BS,Iadecola C,Bulloch K
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Thu Jul 01 00:00:00 EDT 2010
Naive tumor-specific CD4(+) T cells differentiated in vivo eradicate established melanoma.
The Journal of experimental medicine
Xie Y,Akpinarli A,Maris C,Hipkiss EL,Lane M,Kwon EK,Muranski P,Restifo NP,Antony PA
16-5931 was used in Flow cytometry/Cell sorting to study the regression of tumours due to naive tumor-specific CD4(+) T cell differentiation.
Mon Mar 15 00:00:00 EDT 2010
Local mesenchymal stem/progenitor cells are a preferential target for initiation of adult soft tissue sarcomas associated with p53 and Rb deficiency.
The American journal of pathology
Choi J,Curtis SJ,Roy DM,Flesken-Nikitin A,Nikitin AY
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Mon Nov 01 00:00:00 EDT 2010
Local mesenchymal stem/progenitor cells are a preferential target for initiation of adult soft tissue sarcomas associated with p53 and Rb deficiency.
The American journal of pathology
Choi J,Curtis SJ,Roy DM,Flesken-Nikitin A,Nikitin AY
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Mon Nov 01 00:00:00 EDT 2010
The presence of professional phagocytes dictates the number of host cells targeted for Yop translocation during infection.
Cellular microbiology
Durand EA,Maldonado-Arocho FJ,Castillo C,Walsh RL,Mecsas J
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Sun Aug 01 00:00:00 EDT 2010
The presence of professional phagocytes dictates the number of host cells targeted for Yop translocation during infection.
Cellular microbiology
Durand EA,Maldonado-Arocho FJ,Castillo C,Walsh RL,Mecsas J
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Sun Aug 01 00:00:00 EDT 2010
The presence of professional phagocytes dictates the number of host cells targeted for Yop translocation during infection.
Cellular microbiology
Durand EA,Maldonado-Arocho FJ,Castillo C,Walsh RL,Mecsas J
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Sun Aug 01 00:00:00 EDT 2010
The presence of professional phagocytes dictates the number of host cells targeted for Yop translocation during infection.
Cellular microbiology
Durand EA,Maldonado-Arocho FJ,Castillo C,Walsh RL,Mecsas J
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Sun Aug 01 00:00:00 EDT 2010
Depletion of Ly6G/Gr-1 leukocytes after spinal cord injury in mice alters wound healing and worsens neurological outcome.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Stirling DP,Liu S,Kubes P,Yong VW
16-5931 was used in Flow cytometry/Cell sorting to further the knowledge of the functions of neuroinflammation in spinal cord injury (SCI) in mice.
Wed Jan 21 00:00:00 EST 2009
Combination of all-trans retinoic acid and a human papillomavirus therapeutic vaccine suppresses the number and function of immature myeloid cells and enhances antitumor immunity.
Cancer science
Song X,Ye D,Liu B,Cui J,Zhao X,Yi L,Liang J,Song J,Zhang Z,Zhao Q
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Sun Feb 01 00:00:00 EST 2009
Cellular expression and antimicrobial function of a phylogenetically conserved novel histone 1x-like protein on mouse cells: a potential new class of pattern recognition receptor.
Journal of leukocyte biology
Evans DL,Connor MA,Moss LD,Lackay S,Leary JH,Krunkosky T,Jaso-Friedmann L
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2009
TLR2 and its co-receptors determine responses of macrophages and dendritic cells to lipoproteins of Mycobacterium tuberculosis.
Cellular immunology
Drage MG,Pecora ND,Hise AG,Febbraio M,Silverstein RL,Golenbock DT,Boom WH,Harding CV
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Fri Jun 12 00:00:00 EDT 2009
Autoregulation of Th1-mediated inflammation by twist1.
The Journal of experimental medicine
Niesner U,Albrecht I,Janke M,Doebis C,Loddenkemper C,Lexberg MH,Eulenburg K,Kreher S,Koeck J,Baumgrass R,Bonhagen K,Kamradt T,Enghard P,Humrich JY,Rutz S,Schulze-Topphoff U,Aktas O,Bartfeld S,Radbruch H,Hegazy AN,Löhning M,Baumgart DC,Duchmann R,Rudwaleit M,Häupl T,Gitelman I,Krenn V,Gruen J,Sieper J,Zeitz M,Wiedenmann B,Zipp F,Hamann A,Janitz M,Scheffold A,Burmester GR,Chang HD,Radbruch A
16-5931-82 was used immunohistochemistry (paraffin) show that twist1 is expressed by activated T helper 1 effector memory cells.
Mon Aug 04 00:00:00 EDT 2008
Host transmission of Salmonella enterica serovar Typhimurium is controlled by virulence factors and indigenous intestinal microbiota.
Infection and immunity
Lawley TD,Bouley DM,Hoy YE,Gerke C,Relman DA,Monack DM
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunofluorescence
Tue Jan 01 00:00:00 EST 2008
Listeria monocytogenes promotes tumor growth via tumor cell toll-like receptor 2 signaling.
Cancer research
Huang B,Zhao J,Shen S,Li H,He KL,Shen GX,Mayer L,Unkeless J,Li D,Yuan Y,Zhang GM,Xiong H,Feng ZH
Tue May 01 00:00:00 EDT 2007
Mouse neutrophilic granulocytes express mRNA encoding the macrophage colony-stimulating factor receptor (CSF-1R) as well as many other macrophage-specific transcripts and can transdifferentiate into macrophages in vitro in response to CSF-1.
Journal of leukocyte biology
Sasmono RT,Ehrnsperger A,Cronau SL,Ravasi T,Kandane R,Hickey MJ,Cook AD,Himes SR,Hamilton JA,Hume DA
Sun Jul 01 00:00:00 EDT 2007
Mouse neutrophilic granulocytes express mRNA encoding the macrophage colony-stimulating factor receptor (CSF-1R) as well as many other macrophage-specific transcripts and can transdifferentiate into macrophages in vitro in response to CSF-1.
Journal of leukocyte biology
Sasmono RT,Ehrnsperger A,Cronau SL,Ravasi T,Kandane R,Hickey MJ,Cook AD,Himes SR,Hamilton JA,Hume DA
Sun Jul 01 00:00:00 EDT 2007
Mouse neutrophilic granulocytes express mRNA encoding the macrophage colony-stimulating factor receptor (CSF-1R) as well as many other macrophage-specific transcripts and can transdifferentiate into macrophages in vitro in response to CSF-1.
Journal of leukocyte biology
Sasmono RT,Ehrnsperger A,Cronau SL,Ravasi T,Kandane R,Hickey MJ,Cook AD,Himes SR,Hamilton JA,Hume DA
Sun Jul 01 00:00:00 EDT 2007
The mouse mammary carcinoma 4T1: characterization of the cellular landscape of primary tumours and metastatic tumour foci.
International journal of experimental pathology
DuPré SA,Redelman D,Hunter KW
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Flow Cytometry
Mon Oct 01 00:00:00 EDT 2007
Hemorrhagic shock-activated neutrophils augment TLR4 signaling-induced TLR2 upregulation in alveolar macrophages: role in hemorrhage-primed lung inflammation.
American journal of physiology. Lung cellular and molecular physiology
Fan J,Li Y,Vodovotz Y,Billiar TR,Wilson MA
Sat Apr 01 00:00:00 EST 2006
Neutralization of Gro alpha and macrophage inflammatory protein-2 attenuates renal ischemia/reperfusion injury.
The American journal of pathology
Miura M,Fu X,Zhang QW,Remick DG,Fairchild RL
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunohistochemistry (Frozen)
Sat Dec 01 00:00:00 EST 2001
Neutralization of Gro alpha and macrophage inflammatory protein-2 attenuates renal ischemia/reperfusion injury.
The American journal of pathology
Miura M,Fu X,Zhang QW,Remick DG,Fairchild RL
Published figure using Ly-6G/Ly-6C monoclonal antibody (Product # 16-5931-82) in Immunohistochemistry (Frozen)
Sat Dec 01 00:00:00 EST 2001