Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CD72.1 (Lyb-2.1), a type II integral membrane glycoprotein and a member of the C-lectin family of cell surface receptors, is a differentiation antigen of B cells and is found in mouse strains expressing the Lyb-2.1 allotype. CD72.1 is the ligand of CD5 which is distributed on all T cells and a small number of B cells. The 10.1.D2 monoclonal antibody blocks binding of CD5 to CD72 on the B cell surface which leads to inhibition of the positive signal resulting from CD5/CD72 pairing. However, 10.1.D2 can itself trigger a positive signal by binding CD72.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: B-cell differentiation antigen CD72; CD72; Ly-32; Lyb-2; Lymphocyte antigen 32
Gene Aliases: Cd72; CD72c; Ly-19; Ly-32; Ly-m19; Ly32; Lyb-2
UniProt ID: (Mouse) P21855
Entrez Gene ID: (Mouse) 12517
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support