Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive control: Human tonsil, spleen, lung
Cellular location: Secreted.
This gene encodes human lysozyme, whose natural substrate is the bacterial cell wall peptidoglycan (cleaving the beta[1-4]glycosidic linkages between N-acetylmuramic acid and N-acetylglucosamine). Lysozyme is one of the anti-microbial agents found in human milk, and is also present in spleen, lung, kidney, white blood cells, plasma, saliva, and tears. Missense mutations in LYZ have been identified in heritable renal amyloidosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1,4-beta-N-acetylmuramidase C; 4-beta-N-acetylmuramidase C; Allergen Gal d IV; c-type lysozyme; dystrophin; egg white lysozyme; Lysozyme C; lysozyme F1; unnamed protein product
Gene Aliases: LYZ; LYZC; LYZF1; LZM
UniProt ID: (Chicken) P00698, (Human) P61626
Entrez Gene ID: (Chicken) 396218, (Human) 4069
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support