Search Thermo Fisher Scientific
MA1-70034 detects Myeloid Lineage in rat samples.
MA1-70034 has been successfully used in flow cytometry, western blot, immunohistochemistry (frozen sections) procedures.
The MA1-70034 immunogen is anemic rat bone marrow.