Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: RGLEKARQQL QEELRQVSGQ LLEERKKRET HEALARRLQK RVLLLTKERD GMRAILGSYD SELTPAEYSP QLTRRMREAE DMVQKVHSHS AEMEAQLSQA LEELGGQKQR ADMLEMELKM LKSQSSSAEQ SFLFSREEAD TLRLKVEELE GERSRLEEEK RMLEAQLERR ALQGDYDQSR TKVLHMSLNP TSVARQRLRE DHSQLQAECE RLRGLLRAME RGGTVPADLE AAAASLPSSK EVAELKKQVE SAELKNQRLK EVFQTKIQEF RKACYTLTGY QIDITTENQY RLTSLYAEHP GDCLIFKATS PSGSKMQLLE TEFSHTVGEL IEVHLRRQDS I; Positive Samples: HeLa, HepG2, MCF7; Cellular Location: Chromosome, Cytoplasm, Nucleus, centromere, centrosome, cytoskeleton, kinetochore, microtubule organizing center, spindle
MAX dimerization protein belongs to a subfamily of MAX-interacting proteins. This protein competes with MYC for binding to MAX to form a sequence-specific DNA-binding complex, acts as a transcriptional repressor (while MYC appears to function as an activator) and is a candidate tumor suppressor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hMAD1; HsMAD1; MAD1 mitotic arrest deficient-like 1; MAD1-like protein 1; Mitotic arrest deficient 1-like protein 1; Mitotic checkpoint MAD1 protein homolog; Mitotic spindle assembly checkpoint protein MAD1; mitotic-arrest deficient 1, yeast, homolog-like 1; MXD1; Tax-binding protein 181; tumor protein p53 inducible protein 9
Gene Aliases: MAD1; MAD1L1; PIG9; TP53I9; TXBP181
UniProt ID: (Human) Q9Y6D9
Entrez Gene ID: (Human) 8379
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support