Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SAEIVAEFF SFGINSILYQ RGIYPSETFT RVQKYGLTLL VTTDLELIKY LNNVVEQLKD WLYKCSVQKL VVVISNIESG EVLERWQFDI ECDKTAKDDS APREKSQKAI QDEIRSVIRQ ITATVTFLPL LEVSCSFDLL IYTDKDLVVP EKWEESGPQF ITNSEEVRLR S (16-185 aa encoded by BC000356)
Tight junction proteins (TJPs) belong to a family of membrane-associated guanylate kinase (MAGUK) homologs that are involved in the organization of epithelial and endothelial intercellular junctions. TJPs bind to the cytoplasmic C termini of junctional transmembrane proteins and link them to the actin cytoskeleton.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HsMAD2; MAD2 (mitotic arrest deficient, yeast, homolog)-like 1; MAD2-like 1; MAD2-like protein 1; MadL1; Mitotic arrest deficient 2-like protein 1; mitotic arrest deficient, yeast, homolog-like 1; Mitotic spindle assembly checkpoint protein MAD2A
Gene Aliases: HSMAD2; MAD2; MAD2L1
UniProt ID: (Human) Q13257
Entrez Gene ID: (Human) 4085
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support