Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
This gene encodes a member of the Myc superfamily of basic helix-loop-helix leucine zipper transcriptional regulators. The encoded protein forms a heterodimer with the cofactor MAX which binds specific E-box DNA motifs in the promoters of target genes and regulates their transcription. Disruption of the MAX-MXD3 complex is associated with uncontrolled cell proliferation and tumorigenesis. Transcript variants of this gene encoding different isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHc13; Class C basic helix-loop-helix protein 13; FLJ35523; Max dimerization protein 3; Max dimerizer 3; Max-associated protein 3; Max-interacting transcriptional repressor MAD3; MGC2383; Myx
Gene Aliases: 4631412E13Rik; BHLHC13; MAD3; MXD3; MYX
UniProt ID: (Human) Q9BW11, (Rat) Q62912, (Mouse) Q80US8
Entrez Gene ID: (Human) 83463, (Rat) 252915, (Mouse) 17121
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support