Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QASPPPSSA ASPAQQLPTS HPGPGPHATA SATAAGGNGS VEDRFSDDQL VSMSVRELNR HLRGFTKDEV IRLKQKRRTL KNRGYAQSCR YKRVQQKHHL ENEKTQLIQQ VEQLKQEVSR LARERDAYKV KCEKLANSGF REAGSTSDSP SSPEFFL (168-323 aa encoded by BC036689 )
The protein encoded by this gene is a basic leucine zipper transcription factor that plays an important role in the regulation of lineage-specific hematopoiesis. The encoded nuclear protein represses ETS1-mediated transcription of erythroid-specific genes in myeloid cells. This gene contains no introns.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Kreisler; Kreisler (maf-related) leucine zipper protein; Kreisler maf-related leucine zipper homolog; Kreisler maf-related leucine zipper homolog (b-maf); Maf-B; MAFB/Kreisler basic region/leucine zipper transcription factor; MGC43127; Segmentation protein Kr; Transcription factor Maf-1; Transcription factor MafB; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog B; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein B; V-maf musculoaponeurotic fibrosarcoma oncogene homolog B
Gene Aliases: b-maf; kr; Kreisler; KRML; Krml1; Maf1; MAFB; MCTO
UniProt ID: (Human) Q9Y5Q3, (Rat) P54842, (Mouse) P54841
Entrez Gene ID: (Human) 9935, (Rat) 54264, (Mouse) 16658
Molecular Function: basic leucine zipper transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support