Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 6
Recommended positive controls: human brain, mouse brain, rat brain.
Predicted reactivity: Mouse (95%), Rat (95%), Bovine (92%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a type I membrane protein and member of the immunoglobulin superfamily. It is thought to be involved in the process of myelination. It is a lectin that binds to sialylated glycoconjugates and mediates certain myelin-neuron cell-cell interactions. Two alternatively spliced transcripts encoding different isoforms have been described for this gene.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1B236; Brain neuron cytoplasmic protein 3; Myelin-associated glycoprotein; S-MAG (C-term.); sialic acid binding Ig-like lectin 4A; Sialic acid-binding Ig-like lectin 4a; sialic acid-binding immunoglobulin-like lectin 4A; Siglec-4a
Gene Aliases: GMA; MAG; S-MAG; SIGLEC-4A; SIGLEC4A; SPG75
UniProt ID: (Human) P20916, (Rat) P07722, (Mouse) P20917
Entrez Gene ID: (Human) 4099, (Rat) 29409, (Mouse) 17136
Molecular Function: immunoglobulin superfamily cell adhesion molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support