Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSLEQRSLH CKPEEALEAQ QEALGLVCVQ AAASSSSPLV LGTLEEVPTA GSTDPPQSPQ GASAFPTTIN FTRQRQPSEG SSSREEEGPS TSCILESLFR AVITKKVADL VGFLLLKYRA REPVTKAEML ESVIKNYKHC FPEIFGKASE SLQLVFGIDV KEADPTGHSY VLVTCLGLSY DGLLGDNQIM PKTGFLIIVL VMIAMEGGHA PEEEIWEELS VMEVYDGREH SAYGEPRKLL TQDLVQEKYL EYRQVPDSDP ARYEFLWGPR ALAETSYVKV LEYVIKVSAR VRFFFPSLRE AALREEEEGV (1-309 aa encoded by BC017555)
Human malignant neoplasms carry rejection antigens that are recognized by the patients' autologous, tumor directed and specific, cytolytic, CD8+ T lymphocyte clones (CTL). An important group of antigens is coded by the MAGE family of genes. It was identified that melanoma associated with antigens (MAAs). Because MAGE-1 is expressed on a significant proportion of human neoplasms of various hisological tyoes (melanoma, brain tumors of glial origin, neuroblastoma, non-small cell lung cancer, breast, gastric, colorectal, ovarian, renal cell carcinomas) and not on normal tissues, the encoded antigen may serve as a marker of early detection and target for cancer immunotherapy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Antigen MZ2-E; Cancer/testis antigen 1.1; cancer/testis antigen family 1, member 1; CT1.1; mage 1; mage-1; MAGE-1 antigen; melanoma antigen family A 1; melanoma antigen family A, 1 (directs expression of antigen MZ2-E); melanoma antigen family A1; melanoma antigen MAGE-1; Melanoma-associated antigen 1; melanoma-associated antigen MZ2-E
Gene Aliases: CT1.1; MAGE1; MAGE1A; MAGEA1
UniProt ID: (Human) P43355
Entrez Gene ID: (Human) 4100
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support