Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: TLPTTINYTL WSQSDEGSSN EEQEGPSTFP DLETSFQVAL SRKMAELVHF LLLKYRAREP FTKAEMLGSV IRNFQDFFPV IFSKASEYLQ LVFGIEVVE
Not known, though may play a role tumor transformation or progression. In vitro promotes cell viability in melanoma cell lines.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer/testis antigen 1.12; cancer/testis antigen family 1, member 12; CT1.12; MAGE-12 antigen; MAGE12F antigen; melanoma antigen family A, 12; melanoma antigen family A12; Melanoma-associated antigen 12
Gene Aliases: CT1.12; MAGE12; MAGEA12
UniProt ID: (Human) P43365
Entrez Gene ID: (Human) 4111
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support