Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MPLEQRSQHC KPEEGLEARG EALGLVGAQA PATEEQQTAS SSSTLVEVTL GEVPAADSPS PPHSPQGASS FSTTINYTLW RQSDEGSSNQ EEEGPRMFPD LESEFQAAIS RKMVELVHFL LLKYQAREPV TKAEMLESVL RNCQDFFPVI FSKASEYLQL VFGIEVVEVV PISHLYILVT CLGLSYDGLL GDNQVMPKTG LLIIVLAIIA IEGDCAPEEK IWEELSMLEV FEGREDSVFA HPRKLLMQDL VQENYLEYRQ VPGSDPACYE FLWGPRALIE TSYVKVLHHT LKIGGEPHIS YPPLHERALR EGEE
MAGEA2 is a member of the MAGEA gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 80% sequence identity between each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are expressed at a high level in a number of tumors of various histologic types, and are silent in normal tissues with the exception of testis and placenta. The MAGEA genes are clustered on chromosome Xq28. They may be implicated in some hereditary disorders, such as dyskeratosis congenita. This gene has two identical copies at different loci.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer/testis antigen 1.2; cancer/testis antigen family 1, member 2; CT1.2; MAGE-2 antigen; melanoma antigen 2; melanoma antigen family A, 2; melanoma antigen family A2; Melanoma-associated antigen 2; MGC131923
Gene Aliases: CT1.2; MAGE2; MAGEA2; MAGEA2A; MAGEA2B
UniProt ID: (Human) P43356
Entrez Gene ID: (Human) 4101
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support