Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
MAGEA4 is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer/testis antigen 1.4; cancer/testis antigen family 1, member 4; CT1.4; MAGE-4 antigen; MAGE-41 antigen; MAGE-X2 antigen; melanoma antigen family A, 4; melanoma antigen family A4; Melanoma-associated antigen 4; MGC21336
Gene Aliases: CT1.4; MAGE-41; MAGE-X2; MAGE4; MAGE4A; MAGE4B; MAGEA4
UniProt ID: (Human) P43358
Entrez Gene ID: (Human) 4103
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support