Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: MCF-7.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene is a member of the MAGEB gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 68% sequence identity to each other. The promoters and first exons of the MAGEB genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. This gene is expressed in testis, and in a significant fraction of tumors of various histological types. The MAGEB genes are clustered on chromosome Xp22-p21.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer/testis antigen 3.4; cancer/testis antigen family 3, member 4; CT3.4; MAGE-B6 antigen; melanoma antigen family B, 6; melanoma antigen family B6; Melanoma-associated antigen B6
Gene Aliases: CT3.4; MAGE-B6; MAGEB6; MAGEB6A
UniProt ID: (Human) Q8N7X4
Entrez Gene ID: (Human) 158809
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support