Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Predicted reactivity: Mouse (89%), Rat (87%), Bovine (92%).
This gene encodes a member of the E subfamily of MAGE (melanoma antigen-encoding gene) gene family. The gene is intronless and the encoded protein has two of the MAGE domains which are characteristic of MAGE family proteins.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hepatocellular carcinoma-associated protein 3; hepatocellular carcinoma-associated protein HCA3; MAGE-E2 antigen; melanoma antigen family E, 2; melanoma antigen family E2; Melanoma-associated antigen E2
Gene Aliases: HCA3; MAGEE2; RGD1564872
UniProt ID: (Human) Q8TD90
Entrez Gene ID: (Human) 139599, (Rat) 302392
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support