Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
Acts as a transcriptional coactivator for NOTCH proteins. Has been shown to amplify NOTCH-induced transcription of HES1. Potentiates activation by NOTCH3 and NOTCH4 more efficiently than MAML1 or MAML3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mam-2; mastermind-like 2; Mastermind-like protein 2; mastermind-like transcriptional coactivator 2
Gene Aliases: KIAA1819; MAM-3; MAM2; MAM3; MAML2; MLL-MAML2
UniProt ID: (Human) Q8IZL2
Entrez Gene ID: (Human) 84441
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support