Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VLFHSSPALQ PAADHKPGPG ARAEDAAEGR ARRREEGAPG DPEAALEDNL ARIRENHERA LREAKETLQK LPEEIQRDIL LEKKKVAQDQ LRDKAPFRGL PPVDFVPPIG VESREPADAA IREKRAKIKE EIEARKGQAN CPGSSSSCEV EIQMMKHAWN NYKGYAWGLN ELKPISKGGH SSSLFGNIKG ATIVDALDTL FIMEMKHEFE EAKSWVEENL DFNVNAEISV FEVNIRFVGG LLSAYYLSGE EIFRKKAVEL GVKLLPAFHT PSGIPWALLN MKRWMCNIPR AMEATI
This gene encodes a class I mammalian Golgi 1,2-mannosidase which is a type II transmembrane protein. This protein catalyzes the hydrolysis of three terminal mannose residues from peptide-bound Man(9)-GlcNAc(2) oligosaccharides and belongs to family 47 of glycosyl hydrolases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-1,2-mannosidase IA; Man(9)-alpha-mannosidase; Man9-mannosidase; mannosidase 1, alpha; Mannosidase alpha class 1A member 1; mannosidase, alpha, class 1A, member 1; Mannosyl-oligosaccharide 1,2-alpha-mannosidase IA; mannosyl-oligosaccharide alpha-1,2-mannosidase; Processing alpha-1,2-mannosidase IA
Gene Aliases: Aa2-166; HUMM3; HUMM9; Man1a; MAN1A1; MAN9; PCR1
UniProt ID: (Human) P33908, (Mouse) P45700
Entrez Gene ID: (Human) 4121, (Mouse) 17155, (Rat) 294410
Molecular Function:
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support