Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VQLAEKLLP AFNTPTGIPW AMVNLKSGVG RNWGWASAGS SILAEFGTLH MEFIHLSYLT GDLTYYKKVM HIRKLLQKMD RPNGLYPNYL NPRTGRWGQY HTSVGGLGDS FYEYLLKAWL MSDKTDHEAR KMYDDAIEAI EKHLIKKSRG GLTFIGEWKN GHLEKKMGHL ACFAGGMFAL GADGSRADKA GHYLELGAEI ARTCHESYDR TALKLGPESF KFDGAVEAVA VRQAEKYYIL RPEVIETYWY LWRFTHDPRY RQWGWEAALA IEKYCRVNGG FSGVKDVYSS TPTHDDVQQS FFLAETLKYL YLLFSGDDLL PLDHWVFNTE AHPLPVLHLA NTTLSGNPAV R (292-641 aa encoded by BC063300)
Alpha-mannosidases function at different stages of N-glycan maturation in mammalian cells. See MAN2A1 (MIM 154582) for general information.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3.2.1.113; Alpha-1,2-mannosidase IB; alpha1,2-mannosidase; Mannosidase alpha class 1A member 2; mannosidase, alpha, class 1A, member 2; Mannosyl-oligosaccharide 1,2-alpha-mannosidase IB; Processing alpha-1,2-mannosidase IB
Gene Aliases: MAN1A2; MAN1B
UniProt ID: (Human) O60476
Entrez Gene ID: (Human) 10905
Molecular Function:
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support