Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: SSKHKRFDLG LEDVLIPHVD AGKGAKNPGV FLIHGPDEHR HREEEERLRN KIRADHEKAL EEAKEKLRKS REEIRAEIQT EKNKVVQEMK IKENKPLPPV PIPNLVGIRG GDPEDNDIRE K; Positive Samples: LO2, A-549, Mouse brain, Mouse liver; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein
Alpha-mannosidases function at different stages of N-glycan maturation in mammalian cells. See MAN2A1 (MIM 154582) for general information.
โ WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3.2.1.113; Alpha-1,2-mannosidase IB; alpha1,2-mannosidase; mannosidase 1, beta; Mannosidase alpha class 1A member 2; mannosidase, alpha, class 1A, member 2; Mannosyl-oligosaccharide 1,2-alpha-mannosidase IB; Processing alpha-1,2-mannosidase IB
Gene Aliases: AI428775; AI528764; AI854422; MAN1A2; MAN1B; PCR2
UniProt ID: (Human) O60476, (Mouse) P39098
Entrez Gene ID: (Human) 10905, (Mouse) 17156
Molecular Function:
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support