Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: AFRLEEEQKM RPEIAGLKPA NPPVLPAPQK ADTDPENLPE ISSQKTQRHI QRGPPHLQIR PPSQDLKDGT QEEATKRQEA PVDPRPEGDP QRTVISWRGA
Involved in glycoprotein quality control targeting of misfolded glycoproteins for degradation. It primarily trims a single alpha-1,2-linked mannose residue from Man9GlcNAc2 to produce Man8GlcNAc2, but at high enzyme concentrations, as found in the ER quality control compartment (ERQC), it further trims the carbohydrates to Man5-6GlcNAc2. Tremblay L.O., Glycobiology 9:1073-1078(1999) [PubMed: 10521544]. Gonzalez D.S., J. Biol. Chem. 274:21375-21386(1999) [PubMed: 10409699]. Clark H.F., Genome Res. 13:2265-2270(2003) [PubMed: 12975309].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3.2.1.113; endoplasmic Reticulum Class I alpha-mannosidase; Endoplasmic reticulum mannosyl-oligosaccharide 1,2-alpha-mannosidase; endoplasmic reticulum mannosyl-oligosaccharide 1,2-alpha-mannosidase 1; ER alpha 1,2-mannosidase; ER alpha-1,2-mannosidase; ER mannosidase 1; ERMan1; Man9GlcNAc2-specific processing alpha-mannosidase; Man9GlcNAc2-specific-processing alpha-mannosidase; Mannosidase alpha class 1B member 1; mannosidase, alpha, class 1B, member 1; RP11-350O14.2
Gene Aliases: ERMAN1; ERManI; MAN1B1; MANA-ER; MRT15; UNQ747/PRO1477
UniProt ID: (Human) Q9UKM7
Entrez Gene ID: (Human) 11253
Molecular Function:
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support