Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLMRKVPGFV PASPWGLRLP QKFLFLLFLS GLVTLCFGAL FLLPHSSRLK RLFLAPRTQQ PGLEVVAEIA GHAPAREQEP PPNPAPAAPA PGEDDPSSWA SPRRRKGGLR RTRPTGPREE ATAARGNSIP ASRPGDEGVP FRFDFNAFRS RLRHPVLGTR ADESQEPQSQ VRAQREKIKE MMQFAWQSYK RYAMGKNELR PLTKDGYEGN MFGGLSGATV IDSLDTLYLM ELKEEFQEAK AWVGESFHLN VSGEASLFEV NIRYIGGLLS AFYLTGEEVF RIKAIRLGEK LLPAFNTPTG IPKGVVSFKS GNWGWATAGS SSILAEFGSL HLEFLHLTEL SGNQVFAEKV RNIRKVLRKI EKPFGLYPNF LSPVSGNWVQ HHVSVGGLGD SFYEYLIKSW LMSGKTDMEA KNMYYEALEA IETYLLNVSP GGLTYIAEWR GGILDHKMGH LACFSGGMIA LGAEDAKEEK RAHYRELAAQ ITKTCHESYA RSDTKLGPEA FWFNSGREAV ATQLSESYYI LRPEVVESYM YLWRQTHNPI YREWGWEVVL ALEKYCRTEA GFSGIQDVYS STPNHDNKQQ SFFLAETLKY LYLLFSEDDL LSLEDWVFNT EAHPLPVNHS DSSGRAWGRH
MAN1C1 gene ontology annotations related to this gene include metabolic process.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1,2-alpha-mannosidase IC; Alpha-1,2-mannosidase IC; HMIC; Mannosidase alpha class 1C member 1; mannosidase, alpha, class 1C, member 1; Mannosyl-oligosaccharide 1,2-alpha-mannosidase IC; Processing alpha-1,2-mannosidase IC
Gene Aliases: HMIC; MAN1A3; MAN1C; MAN1C1; pp6318
UniProt ID: (Human) Q9NR34
Entrez Gene ID: (Human) 57134
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support