Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLMRKVPGFV PASPWGLRLP QKFLFLLFLS GLVTLCFGAL FLLPHSSRLK RLFLAPRTQQ PGLEVVAEIA GHAPAREQEP PPNPAPAAPA PGEDDPSSWA SPRRRKGGLR RTRPTGPREE ATAARGNSIP ASRPGDEGVP FRFDFNAFRS RLRHPVLGTR ADESQEPQSQ VRAQREKIKE MMQFAWQSYK RYAMGKNELR PLTKDGYEGN MFGGLSGATV IDSLDTLYLM ELKEEFQEAK AWVGESFHLN VSGEASLFEV NIRYIGGLLS AFYLTGEEVF RIKAIRLGEK LLPAFNTPTG IPKGVVSFKS GNWGWATAGS SSILAEFGSL HLEFLHLTEL SGNQVFAEKV RNIRKVLRKI EKPFGLYPNF LSPVSGNWVQ HHVSVGGLGD SFYEYLIKSW LMSGKTDMEA KNMYYEALEA IETYLLNVSP GGLTYIAEWR GGILDHKMGH LACFSGGMIA LGAEDAKEEK RAHYRELAAQ ITKTCHESYA RSDTKLGPEA FWFNSGREAV ATQLSESYYI LRPEVVESYM YLWRQTHNPI YREWGWEVVL ALEKYCRTEA GFSGIQDVYS STPNHDNKQQ SFFLAETLKY LYLLFSEDDL LSLEDWVFNT EAHPLPVNHS DSSGRAWGRH
MAN1C1 gene ontology annotations related to this gene include metabolic process.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1,2-alpha-mannosidase IC; Alpha-1,2-mannosidase IC; HMIC; Mannosidase alpha class 1C member 1; mannosidase, alpha, class 1C, member 1; Mannosyl-oligosaccharide 1,2-alpha-mannosidase IC; Processing alpha-1,2-mannosidase IC
Gene Aliases: HMIC; MAN1A3; MAN1C; MAN1C1; pp6318
UniProt ID: (Human) Q9NR34
Entrez Gene ID: (Human) 57134
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support