Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: VTEIRQYFY RNMTAQNYTY AIRSRLTHVP QGHDGELLCH RIEQEYQAGP LELNREAVLR TSTNLNSQQV IYSDNNGYQM QRRPYVSYVN NSIARNYYPM VQSAFMEDGK SRLVLLSERA HGISSQGNGQ VEVMLHRRLW NNFDWDLGYN LTLNDTSVVH PVLWLLLGSW SLTTALRQRS ALALQHRPVV LFGDLAGTAP KLPGPQQQEA VTLPPNLHLQ ILSIPGWRYS SNHTEHSQNL RKGHRGEAQA DLRRVLLRLY HLYEVGEDPV LSQPVTVNLE AVLQALGSVV AVEERSLTGT WDLSMLHRWS WRTGPGRHRG DTTSPSRPPG GPIITVHPKE IRTFFIHFQQ Q (660-1009 aa encoded by BC033307)
The MAN2B2 (Mannosidase Alpha Class 2B Member 2) gene encodes an enzyme that is involved in the processing and modification of glycoproteins by hydrolyzing alpha-mannosidic linkages. Located on chromosome 4, the MAN2B2 protein is crucial for proper lysosomal function and is part of the glycosyl hydrolase family. Pathogenic variants in the MAN2B2 gene have been linked to a rare congenital disorder of glycosylation (CDG), known as MAN2B2-CDG, characterized by immune deficiency, developmental delay, stroke, and dysmorphic facial features. These mutations lead to an altered mannosylation profile, impacting glycoprotein processing and cellular functions. Studies have shown that reintroducing wild-type MAN2B2 can restore the normal mannosylation profile in patient cells, highlighting its therapeutic potential. Furthermore, recent research has identified circular RNA from this gene (circMAN2B2) that is implicated in the proliferation of hepatocellular carcinoma cells, suggesting a role in cancer biology.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 135 kD alpha-D-mannosidase; 135 kDa alpha-D-mannosidase; Alpha-1,6-mannosidase; core-specific lysosomal alpha-1,6-Mannosidase; Epididymis-specific alpha-mannosidase; HMG17L3; mannosidase 2, alpha B2; Mannosidase alpha class 2B member 2; mannosidase, alpha, class 2B, member 2; NHC
Gene Aliases: Ab2-450; KIAA0935; MAN2B2; mKIAA0935
UniProt ID: (Human) Q9Y2E5, (Mouse) O54782
Entrez Gene ID: (Human) 23324, (Mouse) 17160, (Rat) 360955
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support