Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSNKCDVVV VGGGISGMAA AKLLHDSGLN VVVLEARDHV GGRTYTLRNQ KVKYVDLGGS YVGPTQNRIL RLAKELGLET YKVNEVERLI HHVKGKSYPF RGPFPPVWNP ITYLDHNNFW RTMDDMGREI QSDAPWKAPL AEEWDNMTMK ELLDKLCWTE SAKQLATLFV NLCVTAETHE VSALWFLWYV KQCGGTTRII STTNGGQERK FVGGSGQVSE RIMDLLGDRV KLERPVIYID QTRENVLVET LNHEMYEAKY VISAIPPTLG MKIHFNPPLP MMRNQMITRV PLGSVIKCIV YYKEPFWRKK DYCGTMIIDG EEAPVAYTLD DT (1-331 aa encoded by BC022494)
MAOB (Monoamine Oxidase B), also called MAO, BRAIN, AMINE OXIDASE (FLAVIN-CONTAINING) B, is a protein that in humans is encoded by the MAOB gene. MAOB is a member of the flavin monoamine oxidase family. And it is mapped on Xp11.3. MAOB catalyzes the oxidative deamination of biogenic and xenobiotic amines and plays an important role in the metabolism of neuroactive and vasoactive amines in the central nervous system and peripheral tissues. This protein preferentially degrades benzylamine and phenylethylamine. Like MAOA, it also degrades dopamine. MAO-B is involved in the breakdown of dopamine, a neurotransmitter implicated in reinforcing and motivating behaviors as well as movement. MAO-B inhibition is, therefore, associated with enhanced activity of dopamine, as well as with decreased production of hydrogen peroxide, a source of reactive oxygen species.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adrenalin oxidase; Amine oxidase [flavin-containing] B; HGNC:6834; MAO b; MAO, brain; MAO, platelet; MAO-B; MGC26382; Monoamine oxidase type B; RP1-201D17__B.1; tyramine oxidase
Gene Aliases: MAOB
UniProt ID: (Human) P27338
Entrez Gene ID: (Human) 4129
Molecular Function: oxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support