Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAAPSGVHLL VRRGSHRIFS SPLNHIYLHK QSSSQQRRNF FFRRQRDISH SIVLPAAVSS AHPVPKHIKK PDYVTTGIVP DWGDSIEVKN EDQIQGLHQA CQLARHVLLL AGKSLKVDMT TEEIDALVHR EIISHNAYPS PLGYGGFPKS VCTSVNNVLC HGIPDSRPLQ DGDIINIDVT VYYNGYHGDT SETFLVGNVD ECGKKLVEVA RRCRDEAIAA CRAGAPFSVI GNTISHITHQ NGFQVCPHFV GHGIGSYFHG HPEIWHHAND SDLPMEEGMA FTIEPIITEG SPEFKVLEDA WTVVSLDNQR SAQFEHTVLI TSRGAQILTK LPHEA
The N-terminal methionine excision pathway is an essential process in which the N-terminal methionine is removed from many proteins, thus facilitating subsequent protein modification. In mitochondria, enzymes that catalyze this reaction are celled methionine aminopeptidases (MetAps, or MAPs; EC 3.4.11.18).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDS of metAP-3 within PCR fragment; MAP 1D; metAP 1D; Methionine aminopeptidase 1D, mitochondrial; Methionyl aminopeptidase type 1D, mitochondrial; Peptidase M 1D
Gene Aliases: MAP 1D; MAP1D; MetAP 1D; METAP1D; Metap1l
UniProt ID: (Human) Q6UB28
Entrez Gene ID: (Human) 254042
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support