Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: TSAGSTDRLP YSKSGNKDGV TKSPEKRSSL PRPSSILPPR RGVSGDRDEN SFSLNSSISS SARRTTRSEP IRRAGKSGTS TPTTPGSTAI TPGTPPSYSS RTPGTPGTPS YPRTPHTPGT PKSAILVPSE KKVAIIRTPP KSPATPKQLR LINQPLPDLK NVKSKIGSTD NIKYQPKGGQ VRILNKKIDF SKVQSRCGSK DNIKHSAGGG NVQIVTKKID LSHVTSKCGS LKNIRHRPGG GRVKIESVKL DFKEKAQAKV GSLDNAHHVP GGGNVKIDSQ KLNFREHAKA RVDHGAEIIT QSPGRSSVAS PRRLSNVSSS GSINLLESPQ LATLAEDVTA ALAKQGL
MAP2 (Microtubule Associated Protein 2) exists in two high molecular weight forms (MAP2a & MAP2b) and a low molecular weight form. The expression of MAP2 is developmentally regulated and its multiple forms arise by alternative splicing of a single gene. MAP2 is involved in microtubule assembly, which is an essential step in neurogenesis. The products of similar genes in rat and mouse are neuron-specific cytoskeletal proteins that are enriched in dendrites, implicating a role in determining and stabilizing dendritic shape during neuron development. MAP2 is useful in studies of neuron structure in normal and malignant brain tissue as well as in degenerative diseases such as Alzheimer's Disease (AD).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AC091465.1; DKFZp686I2148; MAP-2; map-2a; map2ab; Microtubule-associated protein 2; OTTHUMP00000163916; OTTHUMP00000208143
Gene Aliases: G1-397-34; MAP-2; MAP2; MAP2A; MAP2B; MAP2C; MAP2R; Mtap-2; Mtap2; repro4
UniProt ID: (Human) P11137, (Mouse) P20357, (Rat) P15146
Entrez Gene ID: (Human) 4133, (Mouse) 17756, (Rat) 25595
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support