Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KIHCASSWI NPDTRDQYLI FGAEEGIYTL NLNELHETSM EQLFPRRCTW LYVMNNCLLS ISGKASQLYS HNLPGLFDYA RQMQKLPVAI PAHKLPDRIL PRKFSVSAKI PETKWCQKCC VVRNPYTGHK YLCGALQTSI VLLEWVEPMQ KFMLIKHIDF PIPCPLRMFE MLVVPEQEYP LVCVGVSRGR DFNQVVRFET VNPNSTSSWF TESDTPQTNV THVTQLERDT ILVCLDCCIK IVNLQGRLKS SRKLSSELTF DFQIESIVCL QDSVLAFWKH GMQGRSFRSN EVTQEISDST RIFRLLGSDR VVVLESRPTD NPTANSNLYI LAGHENSY (537-873 aa encoded by BC071579)
MAP4K3 (also known as GLK) is a member of the STE20 family of serine/threonine protein kinases and contains N-terminal catalytic domain and C-terminal regulatory domain. MAP4K3 is upstream of the MEKK1 target and is stimulated by UV radiation and TNF-alpha and can lead to the activation of the JNK signaling pathway. MAP4K3 can directly activate PKC-T, during TCR signaling and people with systemic lupus erythematosus show considerable enhanced MAP4K3 expression and activation of PKC-T, and the kinase IKK in T cells. MAP4K3 can act as a pro-apoptotic kinase which orchestrates activation of BAX via the concerted posttranscriptional modulation of PUMA, BAD, and BIM. The kinase activity of the encoded protein can be stimulated by UV radiation and tumor necrosis factor-alpha. The protein specifically activates the c-Jun N-terminal kinase (JNK) signaling pathway. Evidence suggests that it functions upstream of mitogen-activated protein kinase kinase 1 (MEKK1). MAP4K3 previously was referred to as RAB8-interacting protein-like 1 (RAB8IPL1), but it has been renamed mitogen-activated protein kinase kinase kinase 3 (MAP4K3).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: germinal center kinase-like kinase; Germinal center kinase-related protein kinase; GLK; MAPK/ERK kinase kinase kinase 3; MEK kinase kinase 3; MEKKK 3; mitogen activated protein kinase kinase kinase kinase 3; Mitogen-activated protein kinase kinase kinase kinase 3
Gene Aliases: 4833416M01Rik; 4833416M07Rik; 9530052P13Rik; GLK; MAP4K3; MAPKKKK3; MEKKK 3; MEKKK3; RAB8IPL1
UniProt ID: (Human) Q8IVH8, (Mouse) Q99JP0, (Rat) Q924I2
Entrez Gene ID: (Human) 8491, (Mouse) 225028, (Rat) 170920
Molecular Function:
voltage-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support