Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is specific to MAP4K4.
MAP4K4 (HGK) is a serine/threonine kinase which plays an important role in stress-activated MAPK core signaling pathways, the response to environmental stress and cytokines such as TNF-alpha. It appears to act upstream of the JUN N-terminal pathway. This protein is thought to interact with the SH3 domain of the adapter proteins Nck. HGK binds, via its CNH regulatory domain, to the N-terminal region of SPG3A. Expression appears to be ubiquitous, expressed in all tissue types examined. Isoform 5 appears to be more abundant in the brain, and isoform 4 is predominant in the liver, skeletal muscle and placenta.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory protein Li 31; FLJ10410; FLJ20373; FLJ90111; hepatocyte progenitor kinase-like/germinal center kinase-like kinase; HPK/GCK-like kinase HGK; MAPK/ERK kinase kinase kinase 4; MEK kinase kinase 4; MEKKK 4; mitogen activated protein kinase kinase kinase kinase 4; Mitogen-activated protein kinase kinase kinase kinase 4; NCK interacting kinase; Nck-interacting kinase; Ste20 group protein kinase HGK
Gene Aliases: 9430080K19Rik; AU043147; AU045934; AW046177; FLH21957; HEL-S-31; HGK; KIAA0687; MAP4K4; MEKKK4; NIK
UniProt ID: (Human) O95819, (Mouse) P97820
Entrez Gene ID: (Human) 9448, (Mouse) 26921, (Rat) 301363
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support